What are the differences in library entry format between MicroSeq ID 16S rDNA 500 Supplemental Library and MicroSEQ ID 16S rDNA 500 Library v2013?
The library entry format for MicroSeq ID 16S rDNA 500 Supplemental Library contains strain information in the Comments column. In addition, the Source column has NCBI information for all entries and the Entry Number column includes accession number for the sequences.