Custom TaqMan™ Small RNA Assay
Applied Biosystems™

Custom TaqMan™ Small RNA Assay

Custom TaqMan Small RNA Assays use TaqMan Real-Time PCR Assays for the analysis of any small RNA molecule.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4398988CNM (750 RT/750 PCR Reactions), Made to Order
4440418CNXS (25 RT/75 PCR Reactions), Made to Order
4398987CNS (50 RT/150 PCR Reactions), Made to Order
4398989CNL (2900 RT/2900 PCR Reactions), Made to Order
Catalog number 4398988CN
Price (USD)
-
Quantity:
M (750 RT/750 PCR Reactions), Made to Order

Custom TaqMan Small RNA Assays use TaqMan Real-Time PCR Assays for the analysis of any small RNA molecule. The novel adaptations in TaqMan Assay design developed for the study of miRNAs are ideal for analysis of any small nucleic acid less than 200 bases long, including newly discovered miRNAs, Piwi-interacting RNA (piRNA), small nuclear RNA (snRNA), and small nucleolar RNA (snoRNA).

  • Flexible and convenient ordering—easy process with our secure Custom TaqMan Small RNA Assay design tool
  • Versatile application—use for the analysis of any small RNA from any species
  • Reliable outcome—pre-formulated assays are optimized to run under universal conditions
  • Powerful miRNA quantitation—the same sensitivity, specificity, and reproducibility as predesigned TaqMan MicroRNA Assays

Sophisticated design pipeline

The miRPipe Small RNA Assay design pipeline is founded on our extensive background and bioinformatics expertise designing TaqMan Gene Expression Assays. To detect very small targets, it includes design features based on empirical data gathered over years of experiments. The resulting automated pipeline includes built-in flexibility features enabling design of assays for the broadest assortment of small RNA sequences. The benefits of TaqMan Assay technology are ideal for analysis of virtually any small RNA, enabling results you can trust from the very first experiment.

Pre-formulated assay

  • Forward primer: 1500 nM
  • Reverse primer: 700 nM
  • FAM-MGB probe: 200 nM
  • RT sequences: 50 nM

Ordering information

First, bioinformatically qualify your sequence target for specificity and sequence quality, then use the Custom TaqMan Small RNA Assay Design Tool to input your sequence and submit it for assay design. This easy-to-use tool is a secure and confidential. If we are unable to design or manufacture an assay for your sequence target of interest, you will not be charged.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions750 Reactions (PCR and RT-PCR)
Product LineTaqMan
Product TypeSmall RNA Assay
QuantityM (750 RT/750 PCR Reactions), Made to Order
Shipping ConditionRoom Temperature
Concentration20X (Assay), 20X (RT Primer)
For Use With (Application)Gene Expression, miRNA analysis
FormLiquid
Unit SizeEach
Contents & Storage
• 1 tube containing RT primer at concentration of 5X (XS and S sizes), 20X (M size), or 60X (L size)
• 1 tube containing a 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.