TaqMan™ Copy Number Assay
Applied Biosystems™

TaqMan™ Copy Number Assay

Applied Biosystems TaqMan Copy Number Assays use gold-standard TaqMan MGB probe chemistry to evaluate the copy number of genomic DNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4400293L (2900 reactions), made to order
4400291S (360 reactions), made to order
4400292M (750 reactions), made to order
Catalog number 4400293
Price (USD)
1,640.65
Online exclusive
1,764.00
Save 123.35
Each
Quantity:
L (2900 reactions), made to order

Applied Biosystems TaqMan Copy Number Assays use gold-standard TaqMan MGB probe chemistry to evaluate the copy number of genomic DNA targets using Applied Biosystems real-time PCR instruments and software. TaqMan Copy Number Assays are run together with a TaqMan Copy Number Reference Assay in a duplex qPCR reaction; the copy number assay detects the target sequence, and the reference assay detects a sequence that is known to be present in two copies in the diploid genome. The results are then analyzed by the relative quantitation method using Applied Biosystems CopyCaller Software.

The predesigned TaqMan Copy Number Assay collection consists of human and mouse assay libraries for copy number variation (CNV) analysis. The human assay collection includes over 1.6 million assays targeting gene exons and introns, extragenic regions, and CNV sequences from the Database of Genomic Variants (DGV). The mouse collection includes over 180,000 assays targeting gene exons. Assays to common vector marker and reporter genes are also available for transgenic studies.

Benefits:

  • Specific—gold-standard TaqMan chemistry with minor groove binder (MGB) probes increase assay specificity by enabling shorter probes
  • Reproducible—robust assay designs developed with our bioinformatics pipeline enable accurate, reproducible, and reliable results
  • Easy to interpret—CopyCaller Software provides the calculated copy number and predicted copy number, along with confidence value and z-score quality metrics
  • Fast and simple—setup to primary analysis in 3–4 hours

Approximate ship time

4–6 days in North America and 6–10 days in Europe

All assay designs are the product of our bioinformatics pipeline, optimized over the course of more than a decade by leveraging manufacturing and assay performance data. TaqMan Assays have been cited in over 40,000 publications and are backed by more than 350 patents.

All of our predesigned TaqMan Assays are covered by the TaqMan Assays qPCR Guarantee.*

TaqMan Copy Number Reference Assays are sold separately.

Recommended master mix (sold separately): TaqMan Genotyping Master Mix (Cat. No. 4371355)

*Terms and conditions apply. For complete details, go to www.thermofisher.com/taqmanguarantee.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
3'Primer ModificationNone
5'Primer ModificationNone
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
Internal Probe ModificationFAM (5'), MGB (Minor Groove Binder) (3')
No. of Reactions2900 reactions
Product LineTaqMan
Product TypeCopy Number Assay
Purification MethodSolid Phase Extraction (SPE)
Purity or Quality GradeRNase-Free, DNase-Free
QuantityL (2900 reactions), made to order
Shipping ConditionRoom Temperature
SpeciesHuman
Concentration60X
Detection MethodPrimer-probe
FormFrozen
Label or DyeFAM
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

Can I use the CopyCaller Software if I am using a non-Applied Biosystems Real-Time PCR instrument?

Yes, although the data will not be directly compatible. You can use the example files that install with the CopyCaller Software as a template to use with your data. You will have to copy/paste in the sample/target names and Ct values before importing into CopyCaller Software.

My data will not import into the CopyCaller Software. What could be wrong?

In the instrument software, make sure that you selected “Results” from the export options. The data need to be set up as duplex reactions as well. If you ran the copy number assay and the reference assay in separate wells, the data will not be in the correct format.

CopyCaller Software gave the error "XXX.txt is not a compatible file and cannot be added to the analysis. Verify that the file format is compatible with the software." What can I do now?

Check your export results with the instrument software. The data columns need to be in the following order: Well, Sample Name, Target Name, Task Reporter, Quencher, CT. If needed, rearrange the columns and export the data again for CopyCaller Software.

My results are different from what I expected when working with your Copy Number Variation Assay. Is there something wrong with the assay?

Check that the reference assay is performing consistently across all samples. Check the concentration of your samples and make sure they are normalized. We also recommend you to use several copy number assays targeting the same variation to validate the data.

What can I do if the copy number confidence values are very low?

Confidence values can be low for several reasons. Check your data for the following:

- Large variability in ΔCt values across the plate.
- Low number of replicates per sample (recommendation is for at least 4 replicates per sample).
- Sample copy number is high (> 3). You may want to use copy number bins instead for high copy numbers. Also keep in mind that CopyCaller Software is best for copy ranges of 1-5. If you have higher copy numbers, such as with transfections/transductions, it is best to use a standard curve.