Product Image
Applied Biosystems™

Megaplex™ Primer Pools, Human Pools A v2.1

Megaplex™ Primer Pools, Human Pools A v2.1 is available under a single part number containing both the Megaplex™ RT PrimersRead more
Have Questions?
Catalog NumberQuantity
440100950 reactions
Catalog number 4401009
Price (USD)
1,880.00
Each
Quantity:
50 reactions
Price (USD)
1,880.00
Each
Megaplex™ Primer Pools, Human Pools A v2.1 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex PreAmp Primers, Human Pools A v2.1. Megaplex™ RT Primers, Human Pool A v2.1 contains RT primers for 377 unique microRNAs and 4 controls. For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be better characterized, more broadly expressed and⁄or expressed at a higher level compared to Human Pool B. Human Pool A is intended for use with the TaqMan® Array Human MicroRNA A Card v2.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions50 Reactions
PrimerRT Primers, PreAmp Primers
Product LineMegaplex, TaqMan
Product TypePrimer Pool
Quantity50 reactions
SpeciesHuman
Sufficient For50 Reactions
For Use With (Application)miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -20°C.