Applied Biosystems® Megaplex™ Primer Pools, Rodent Pool A is available under a single part number containing both the Megaplex™ RTRead more
Have Questions?
Catalog Number
Quantity
4401090
50 reactions
Catalog number 4401090
Price (USD)
1,992.00
Each
Quantity:
50 reactions
Price (USD)
1,992.00
Each
Applied Biosystems® Megaplex™ Primer Pools, Rodent Pool A is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Rodent Pools A. Megaplex™ RT Primers, Rodent Pools A contains RT primers for 335 and 226 unique microRNAs for mouse and rat respectively plus 4 species specific controls. For use when assay sensitivity of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be better characterized, more broadly expressed and⁄or expressed at a higher level compared to Rodent Pool B. Rodent Pool A is intended for use with the TaqMan® Rodent MicroRNA A Array v2.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)10 X
No. of Reactions50 Reactions
PrimerRT Primers, PreAmp Primers
Product LineMegaplex, TaqMan
Product TypePrimer Pool
Quantity50 reactions
SpeciesRat, Mouse
Sufficient For50 Reactions
Concentration10X
For Use With (Application)miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Contains 150μL of 10X primers (Megaplex™ PreAmp Primers, Rodent Pool A) and 50μL of 10X primer (Megaplex™ RT Primers, Rodent Pool A). Store at -15 to -25°C.