MicroSEQ&trade; <i>Salmonella</i> spp. Detection Kit
Applied Biosystems™

MicroSEQ™ Salmonella spp. Detection Kit

This kit is recommended for users who have already purchased the starter kit but would like to perform more tests.Read more
Have Questions?
Catalog NumberQuantity
44039301 kit
Catalog number 4403930
Price (USD)
-
Quantity:
1 kit
This kit is recommended for users who have already purchased the starter kit but would like to perform more tests. The MicroSEQ™ Salmonella spp. Detection Kit detects Salmonella spp. simply, reliably, and rapidly in food samples. The assay uses the polymerase chain reaction (PCR) to amplify a unique microorganism specific DNA target sequence and a TaqMan™ probe to detect the amplified sequence.
• Enjoy ease-of-use from a streamlined, lypholized format and a simple, software guided 3-step process
• Get your results quickly with time to results of less than 3 hours after sample enrichment (less than 19 hours total)
• Detect 1-3 colony-forming units (CFU) per 25 grams of food
• Get optimal performance on a wide range of food sample types
• Eliminate false negatives with internal positive control
• Validated by the AOAC Research Institute and AFNOR according to ISO 16140


Lyophilized for Efficiency and Ease of Use
For maximum ease of use, and consistency of results, the reagents used in the MicroSEQ™ Salmonella spp. Detection Kit are lyophilized into preformatted assay beads. The beads hold the active enzyme, the target-specific primer and probe set, Internal Positive Control (IPC) and other reagents for PCR. The IPC is provided to help eliminate false negatives by detecting the presence of materials that can inhibit target amplification.

Simple, Three-Step Procedure
There is minimal training or prior experience needed for you to perform the assay in these three simple steps.
1. Enrich sample
2. Prepare DNA
3. Run test

AOAC and AFNOR Certified
The full workflow including the MicroSEQ™ Salmonella spp. Detection Kit, PrepSEQ sample preparation kits, 7500 Fast instrument and RapidFinder™ Express software has earned the Performance Tested Methods certification from the AOAC Research Institute, and AFNOR validation status.

Optimized Sample Preparation
The MicroSEQ™ Salmonella spp. Detection Kit is optimized for use with PrepSEQ™ Sample Preparation Kits. PrepSEQ kits reduce pre-enrichment time, enhance assay performance and enable you to enrich and prepare high quality samples from a broad range of food sample types for detection of 1-3 cfu per 25 grams of sample.

Clear Results using a Software-Guided Procedure
After sample enrichment and preparation, RapidFinder™ Express Software guides you through the entire workflow with on-screen instructions. Amplification, detection, data collection and analysis are fully automated. When the assay is complete, the software presents an easy-to-read screen that allows you to view the results in each reaction tube. Results are clearly displayed and can be labeled with flags, notifications and prompts that enable you to quickly interpret the data and take appropriate action.

A Complete Solution
The MicroSEQ Salmonella spp. Detection Kit is part of a complete food testing solution. The kit includes everything required to run 96 reactions. All components have been designed for rapid implementation in food testing laboratories and performance-verified to make Salmonella detection as fast, easy and reliable as possible.

For Research Use Only. Not for use in diagnostics procedures.
Specifications
For Use With (Equipment)7500 Fast System, StepOne™, Fast Mode, StepOnePlus™, StepOnePlus™, Fast Mode
Hot StartBuilt-In Hot Start
Kit TypeMicroSEQ Salmonella spp. Detection Kit
No. of Reactions96 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Fast DNA Polymerase
Product LineMicroSEQ
Quantity1 kit
Sample TypeAny Swab Type, Food, Water Samples
Target Organism ClassSalmonella spp.
Detection MethodPrimer-probe
For Use With (Application)Food and Beverage
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
This kit includes 12 8-tube strips containing assay beads (store at 2 to 8°C in the dark), optical 8-tube strips and 8-cap strips (store at 22 to 28°C), and a Negative Control (store at 2 to 8°C).

These reagents are sufficient for 96 reactions.

Frequently asked questions (FAQs)

Which sample preparation kit is compatible with the MicroSEQ Salmonella spp. Detection Kit?

The MicroSEQ Salmonella spp. Detection Kit is optimized for use with PrepSEQ Sample Preparation Kits.

Citations & References (1)

Citations & References
Abstract
Rapid detection of Salmonella in pet food: design and evaluation of integrated methods based on real-time PCR detection.
Authors:Balachandran P, Friberg M, Vanlandingham V, Kozak K, Manolis A, Brevnov M, Crowley E, Bird P, Goins D, Furtado MR, Petrauskene OV, Tebbs RS, Charbonneau D,
Journal:J Food Prot
PubMed ID:22289596
Reducing the risk of Salmonella contamination in pet food is critical for both companion animals and humans, and its importance is reflected by the substantial increase in the demand for pathogen testing. Accurate and rapid detection of foodborne pathogens improves food safety, protects the public health, and benefits food producers ... More