Which sample preparation kit is compatible with the MicroSEQ Salmonella spp. Detection Kit?
The MicroSEQ Salmonella spp. Detection Kit is optimized for use with PrepSEQ Sample Preparation Kits.
Search
View additional product information for MicroSEQ™ Salmonella spp. Detection Kit - FAQs (4403930)
1 product FAQs found