TaqMan™ Newcastle Disease Virus (NDV) and Xeno™ RNA Controls
Applied Biosystems™

TaqMan™ Newcastle Disease Virus (NDV) and Xeno™ RNA Controls

TaqMan™ NDV and Xeno™ RNA Controls are synthetic positive controls for one-step, real-time, RT-PCR amplification of Newcastle Disease Virus RNA and Xeno™ RNA Internal Positive Control.
Have Questions?
Catalog NumberQuantity
44068751 kit
Catalog number 4406875
Price (USD)
129.00
Each
Quantity:
1 kit
Price (USD)
129.00
Each

TaqMan™ NDV and Xeno™ RNA Controls are synthetic positive controls for one-step, real-time, RT-PCR amplification of Newcastle Disease Virus RNA and Xeno™ RNA Internal Positive Control.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
FormatTube(s)
No. of Reactions15 Reactions
Product LineTaqMan, Xeno
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity1 kit
Shipping ConditionDry Ice
SpeciesAvian
Target Organism ClassNewcastle Disease Virus (NDV)
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Store at -20°C. Includes NDV Control RNA, Xeno™ RNA Control, Nucleic Acid Dilution Solution.