Product Image
Applied Biosystems™

TaqMan™ Array Human Signaling in Gap Junctions

These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or詳細を見る
製品番号(カタログ番号)数量
44141821 standard plate
製品番号(カタログ番号) 4414182
価格(JPY)
27,500
Each
数量:
1 standard plate
These 96-well plates are pre-configured with the most appropriate TaqMan® Gene Expression Assays for a specific biological process, pathway, or disease state. Each plate contains predefined assays and endogenous controls dried-down in the wells, ready for accurate assessment of an entire gene signature in one simple experiment. The TaqMan® Array Human Signaling In Gap Junctions 96-well Plate contains 92 assays to genes associated with signaling in gap junctions and 4 assays to candidate endogenous control genes.
研究用途にのみご使用ください。診断目的には使用できません。
仕様
フォーマット96-well plate
内部プローブの変更MGB(マイナーグルーブバインダー)(3')、非蛍光クエンチャー(3')、FAM(5')
反応数1 x 20 μL Reactions
製品ラインTaqMan
数量1 standard plate
研究カテゴリーCell Cycle & Proliferation, Biomarker-Related Pathway(s), Signal Transduction, Extracellular Matrix & Cell Adhesion
Human
形状乾燥
標識または色素FAM
反応速度スタンダード
Unit SizeEach
組成および保存条件
Each TaqMan™ Array Plate well contains a single TaqMan™ Assay for one 20 μL reaction. The plates can be stored at room temperature or at 4°C.

よくあるご質問(FAQ)

What is the minimum amount of TaqMan array plates I need to purchase to submit an order?

The minimum amount of TaqMan Array plates needed to submit an order is based on the plate type:
• Inventoried TaqMan array plates: 1 plate
• TaqMan Flexible Content panel array plates: 6 plates
• Custom TaqMan array plates: 6 plates
• Specialty TaqMan array plates: 10-20 plates (depends on assay format and configuration). Please contact our Specialty Plates team at Specialty_Plates@thermofisher.com for details.

Can our TaqMan Array Plates be run on non-Applied Biosystem real-time PCR instruments?

We recommend using our Plastics Selection Tool to verify which plates are compatible with your instrument model, https://www.thermofisher.com/us/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html.

Based on our testing, the 96-well TaqMan Array Plates can be used on the following non-Applied Biosystem real-time PCR instruments:
• Standard (0.2mL) 96-well Plates: Bio-Rad IQ5
• Fast (0.1mL) Plates: Bio-Rad CFX Connect and Bio-Rad CFX96 Touch

How do I obtain TaqMan documentation files for my TaqMan array plates?

Beginning in April 2016, TaqMan documentation files for TaqMan Array Plates (assay information files (AIF), experimental setup files, and plate layout files) can be downloaded from the Thermo Fisher Scientific website at https://www.thermofisher.com/taqmanfiles. To obtain your TaqMan documentation files:
• Select the appropriate product type.
• Enter the information being requested for the product type.
• Fixed TaqMan array plates: Enter the Part number and Batch number
• TaqMan Flexible Content panel array plates/Custom TaqMan array plates: Enter the Sales Order number and Batch number
• Click Submit.