VetMAX&trade; <i>M. hyopneumoniae</i> Reagents, Applied Biosystems&trade; design
Applied Biosystems™

VetMAX™ M. hyopneumoniae Reagents, Applied Biosystems™ design

VetMAX™ M. hyopneumoniae Reagents Applied Biosystems Design is a TaqMan™ primer probe mix for real-time PCR amplification of Mycoplasma hyopneumoniae DNA and Xeno™ DNA Internal Positive Control.
Have Questions?
Catalog NumberQuantity
4415217100 reactions
Catalog number 4415217
Price (USD)
594.00
Each
Quantity:
100 reactions
Price (USD)
594.00
Each

VetMAX™ M. hyopneumoniae Reagents Applied Biosystems Design is a TaqMan™ primer probe mix for real-time PCR amplification of Mycoplasma hyopneumoniae DNA and Xeno™ DNA Internal Positive Control.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
No. of Reactions100 Reactions
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity100 reactions
Shipping ConditionDry Ice
SpeciesSwine/Porcine
Sufficient For100 Reactions
Target Organism ClassM. hyopneumoniae
Unit SizeEach
Contents & Storage
Store at -20°C. Includes Mhyo AB Design TaqMan™ Primer Probe mix.