VetMAX™-Plus qPCR Master Mix
Applied Biosystems™

VetMAX™-Plus qPCR Master Mix

For use with VetMAX™ Reagents and VetMAX™ Controls. The VetMAX™-Plus qPCR Master Mix is designed for multiplex amplification of animalRead more
Have Questions?
Catalog NumberQuantity
4415327100 Reactions
Catalog number 4415327
Price (USD)
437.65
Online exclusive
486.00
Save 48.35
Each
Quantity:
100 Reactions
Price (USD)
437.65
Online exclusive
486.00
Save 48.35
Each
For use with VetMAX™ Reagents and VetMAX™ Controls. The VetMAX™-Plus qPCR Master Mix is designed for multiplex amplification of animal pathogen DNA targets using a rapid qPCR strategy. The kit includes Xeno™ DNA Control, which serves as an internal positive control for DNA purification and qPCR. DNA is amplified using AmpliTaq Gold® DNA Polymerase on a real-time PCR system.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System
Hot StartBuilt-In Hot Start
No. of Reactions100 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineVetMAX
Product TypePCR Reagents and Detection Kit for Animal Health
Quantity100 Reactions
Sample TypeDNA
Shipping ConditionDry Ice
Sufficient For100 Reactions
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
FormLiquid
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Store at -20°C. Includes 100 reactions with the following components: qPCR Master Mix, Xeno™ DNA Control, and Nuclease-free water.