Boc-L-serine, 99%, anhydrous
Boc-L-serine, 99%, anhydrous
Boc-L-serine, 99%, anhydrous
Thermo Scientific Chemicals

Boc-L-serine, 99%, anhydrous

CAS: 3262-72-4 | C8H15NO5 | 205.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 442560050
Price (USD)
49.65
Online exclusive
55.40
Save 5.75
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
49.65
Online exclusive
55.40
Save 5.75
Each
Chemical Identifiers
CAS3262-72-4
IUPAC Name(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-hydroxypropanoic acid
Molecular FormulaC8H15NO5
InChI KeyFHOAKXBXYSJBGX-MLHKIVSYNA-N
SMILESCC(C)(C)OC(=O)N[C@@H](CO)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH98.5 to 101.5 %
Water=<1 %
Specific optical rotation-6.5° to -8.5° (20°C, 589 nm) (c=2, water)
Appearance (Color)White
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only