OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700 - FAQs
OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700 - FAQs
View additional product information for OpenArray™ Plate Adaptor for Dual Flat Block GeneAmp™ PCR System 9700 - FAQs (4426824)
1 product FAQs found
I have an Applied Biosystems GeneAmp PCR System 9700. What additional accessories do I need to perform off-line cycling for Applied Biosystems QuantStudio OpenArray plates?
You need an Applied Biosystems OpenArray Plate Adaptor (Part Number 4426824). You do not need a compression pad.