TaqMan™ Pri-miRNA Assay
Applied Biosystems™

TaqMan™ Pri-miRNA Assay

TaqMan Pri-miRNA Assays quantitate primary microRNA (pri-miRNA) transcripts, the most upstream RNA molecules in the microRNA biogenesis pathway.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4427013M (750 reactions), made to order
4427012S (360 reactions), made to order
4427014L (2900 reactions), made to order
Catalog number 4427013
Price (USD)
779.20
Special offer
Online exclusive
Ends: 11-Dec-2026
974.00
Save 194.80
Each
Quantity:
M (750 reactions), made to order

TaqMan Pri-miRNA Assays quantitate primary microRNA (pri-miRNA) transcripts, the most upstream RNA molecules in the microRNA biogenesis pathway. They are designed using our state-of-the-art bioinformatics pipeline, delivering gold-standard assay performance and data quality for pri-miRNA studies.

  • Highly specific – quantitiate miRNA transcription from a single genomic locus
  • Fast, simple, and scalable – two-step RT-qPCR provides high-quality results
  • A new perspective – measure miRNA expression at the gene level

TaqMan Pri-miRNA Assays apply the same high sensitivity, superior specificity, and wide linear dynamic range for which all TaqMan genomic assays are renowned to the detection of pri-miRNA transcripts. As a result of their high sensitivity, TaqMan Pri-miRNA Assays require as little as 1 ng of sample input to quantify expression from moderate- to high-expression miRNA loci.

Design and selection
Since all pri-miRNA transcripts have not been mapped, these assays are designed within close proximity to each stem-loop sequence represented in the Sanger miRBase sequence repository. This ensures accurate measurement of the transcript of interest. Each publicly available stem-loop can be matched to a TaqMan Pri-miRNA Assay and TaqMan MicroRNA Assay, enabling the RNA sequences produced at either end of the miRNA maturation pathway to be quantified independently.

Pre-formulated assay (20X; 60X mix)

  • 2 unlabeled PCR primers (900 nM each final 1X reaction concentration)
  • 1 FAM dye-labeled TaqMan MGB probe (250 nM final 1X reaction concentration)

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Fidelity (vs. Taq)60 X
FormatTube
No. of Reactions750 reactions
Product LineTaqMan
Product TypePri-miRNA Assay
QuantityM (750 reactions), made to order
Shipping ConditionRoom Temperature
Sufficient For750 Reactions
Concentration20X
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing a 20X (S and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

Can a TaqMan Pri-miRNA Assay detect more than a single stem-loop?

TaqMan Pri-miRNA Assays are designed to detect the transcript that contains the targeted stem loop. In some instances, e.g. when stem-loop sequences are clustered together and are transcribed on the same transcript, a TaqMan Pri-miRNA Assay targeting one of the stem-loops will also detect the other stem loops in the cluster.

Do the TaqMan MicroRNA Assays detect only the mature form of a miRNA? Vice versa, do the TaqMan Pri-miRNA Assays detect only the pri-form?

Yes. The specific stem-loop chemistry of the TaqMan MicroRNA Assay and its design prevent amplification of the pri-form. Vice versa, the chemistry of the TaqMan Pri-miRNA Assays is not amenable to amplify the mature form. In particular cases, assays designed to detect mature miRNAs derived from the 3' arm of the stem loop may also detect pre-miRNA.

What endogenous control to use for pri-miRNA?

TaqMan endogenous control assays are compatible for pri-miRNA (visit the TaqMan Endogenous Control Assays web portal for more information).

Which kit should I use for the reverse transcription step for TaqMan Pri-miRNA assays (Cat. No. 4427012)?

We recommend the following kits:

  • High-Capacity cDNA Reverse Transcription Kit (Cat. No. 4374966).
  • High-Capacity RNA-to-cDNA Kit (Cat. No. 4387406).


Find additional tips, troubleshooting help, and resources within our PCR and cDNA Synthesis Support Center.