Path-ID™ Multiplex One-Step Kit
Path-ID™ Multiplex One-Step Kit
Path-ID™ Multiplex One-Step Kit
Path-ID™ Multiplex One-Step Kit
Path-ID™ Multiplex One-Step Kit
Path-ID™ Multiplex One-Step Kit
Applied Biosystems™

Path-ID™ Multiplex One-Step Kit

The Path-ID™ Multiplex One-Step RT-PCR Kit is designed for multiplex, quantitative, reverse transcription PCR (qRT-PCR). It is optimized for theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
4428207500 Reactions
4428206100 Reactions
44400221000 Reactions
Catalog number 4428207
Price (USD)
1,804.65
Online exclusive
1,880.00
Save 75.35
Each
No. of Reactions:
500 Reactions
Price (USD)
1,804.65
Online exclusive
1,880.00
Save 75.35
Each
The Path-ID™ Multiplex One-Step RT-PCR Kit is designed for multiplex, quantitative, reverse transcription PCR (qRT-PCR). It is optimized for the amplification of up to four targets simultaneously using your RNA samples and TaqMan® primer⁄probe sets. Reactions are run using a single-tube, one-step procedure to reverse-transcribe the RNA and amplify your targets. The kit contains a Multiplex Enzyme Mix with Arrayscript™ reverse transcriptase and AmpliTaq Gold® DNA Polymerase, and a buffer mix.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7500 Fast System, 7500 System, 7700 System, 7900HT System, AB 7900 HT 384-Well System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode
FormatTube(s)
Hot StartBuilt-In Hot Start
No. of Reactions500 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LinePath-ID
Product TypeOne-Step RT-PCR Kit
Quantity500 reactions
Reverse TranscriptaseArrayScript™
Sample TypeTotal RNA, Viral RNA, mRNA
Sufficient For500 Reactions
Detection MethodPrimer-probe
For Use With (Application)Pathogen Detection, Virus Detection
FormLiquid
PCR Method1-step RT-qPCR, Multiplex qPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
•7 mL of 2X Multiplex RT-PCR Buffer
• 1.4 mL of 10X Multiplex Enzyme Mix including AmpliTaq Gold and ArrayScript
• 25 mL of Nuclease-free water
• Store the kit at -20°C; the Nuclease-free water may be stored at -20°C, 4°C, or room temperature

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.