TaqMan™ Universal Master Mix II, with UNG, 2 x 5 mL - FAQs
TaqMan™ Universal Master Mix II, with UNG, 2 x 5 mL - FAQs
View additional product information for TaqMan™ Universal Master Mix II, with UNG - FAQs (4440042, 4440046, 4440045, 4440044, 4440039, 4440038)
2 product FAQs found
Do I have to use the Universal Master Mix with my TaqMan MicroRNA Assays? If so, which one?
Yes. This is the recommended master mix for TaqMan MicroRNA Assays. You can use Universal Master Mix or Universal Master Mix II (with or without UNG). You can also use the Fast Advanced Master Mix.
Is the 2X Environmental Master Mix the same as the TaqMan Universal PCR Master Mix, with or without UNG?
No, the 2X Environmental Master Mix (EMM) is not the same as the TaqMan Universal PCR Master Mix (with or without AmpErase UNG). The 2X Environmental Master Mix is specifically formulated to detect specific bacterial pathogens with greater specificity and sensitivity. It includes AmpliTaq Gold DNA Polymerase, LD, which is a highly purified version of AmpliTaq Gold DNA Polymerase. Every effort has been made in the development of this new polymerase to eliminate bacterial DNA from the enzyme preparation.