TaqMan™ Universal Master Mix II, with UNG
Applied Biosystems™

TaqMan™ Universal Master Mix II, with UNG

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to performRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44400442 x 5 mL
44400421 x 1 mL
44400381 x 5 mL
44400455 x 5 mL
444004610 x 5 mL
44400391 x 50 mL
Catalog number 4440044
Price (USD)
1,079.52
Special offer
Online exclusive
Ends: 11-Dec-2026
1,384.00
Save 304.48
Each
Quantity:
2 x 5 mL
Price (USD)
1,079.52
Special offer
Online exclusive
Ends: 11-Dec-2026
1,384.00
Save 304.48
Each

TaqMan Universal Master Mix II is a convenient mix of components (except primers, probes, template, and water) necessary to perform a real-time polymerase chain reaction (PCR). Use TaqMan Universal Master Mix II to amplify complementary DNA (cDNA) and DNA targets for a variety of applications, including quantitation and genotyping.

This TaqMan Universal Master Mix II is supplied as a 2X concentrate and contains:
• AmpliTaq Gold DNA Polymerase, UP (Ultra Pure)
• dNTPs (with dUTP)
• ROX Passive Reference
• Uracil-N-glycosylase (UNG)
• Optimized buffer components

Note: This TaqMan Universal Master Mix II contains uracil-N-glycoslyase (UNG). See a complete listing of all TaqMan Universal Master Mix II products, with and without UNG.

TaqMan Universal Master Mix II has been optimized for use with primers and TaqMan probes that have been designed according to Thermo Fisher Scientific guidelines. The master mix can be used with custom TaqMan assays available from our custom assay service, or with pre-optimized assays such as:
• TaqMan gene expression assays
• TaqMan microRNA assays
• TaqMan drug metabolism genotyping assays
• TaqMan SNP genotyping assays

For RNA quantitation experiments, TaqMan Universal Master Mix II is used in the second step of a two-step reverse transcription/polymerase chain reaction (RT-PCR) protocol. The cDNA template used with the master mix can be generated in a reverse transcription reaction using cDNA synthesis kits available from Thermo Fisher Scientific.

Alternative product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of TaqMan Universal Master Mix II and added additional capabilities for your gene expression analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)2 X
For Use With (Equipment)7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineTaqMan
Product TypeUniversal Master Mix II
Purity or Quality GradeUP (Ultra Pure)
Quantity2 x 5 mL
Sample TypeDNA (Genomic), cDNA
Volume10 mL
Concentration2X
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Genotyping, miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C in the dark.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

Frequently asked questions (FAQs)

Do I have to use the Universal Master Mix with my TaqMan MicroRNA Assays? If so, which one?

Yes. This is the recommended master mix for TaqMan MicroRNA Assays. You can use Universal Master Mix or Universal Master Mix II (with or without UNG). You can also use the Fast Advanced Master Mix.

Is the 2X Environmental Master Mix the same as the TaqMan Universal PCR Master Mix, with or without UNG?

No, the 2X Environmental Master Mix (EMM) is not the same as the TaqMan Universal PCR Master Mix (with or without AmpErase UNG). The 2X Environmental Master Mix is specifically formulated to detect specific bacterial pathogens with greater specificity and sensitivity. It includes AmpliTaq Gold DNA Polymerase, LD, which is a highly purified version of AmpliTaq Gold DNA Polymerase. Every effort has been made in the development of this new polymerase to eliminate bacterial DNA from the enzyme preparation.