TaqMan™ Universal Master Mix II, no UNG, 10 x 5 mL - FAQs
TaqMan™ Universal Master Mix II, no UNG, 10 x 5 mL - FAQs
View additional product information for TaqMan™ Universal Master Mix II, no UNG - FAQs (4440043, 4440041, 4440040, 4440047, 4440049, 4440048)
1 product FAQs found