Product Image
Applied Biosystems™

TaqMan™ MicroRNA Assay

Applied Biosystems TaqMan MicroRNA Assays quantitate miRNAs with the specificity and sensitivity of TaqMan Assay chemistry. A simple, two-step protocolRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4440885XS (25 RT/75 PCR reactions), Made to Order
4427975S (50 RT/150 PCR reactions), Inventoried
4440887M (750 RT/750 PCR reactions), Made to Order
4440886S (50 RT/150 PCR reactions), Made to Order
4440888L (2900 RT/2900 PCR reactions), Made to Order
Catalog number 4440885
Price (USD)
251.65
Online exclusive
271.00
Save 19.35
Each
Quantity:
XS (25 RT/75 PCR reactions), Made to Order

Applied Biosystems TaqMan MicroRNA Assays quantitate miRNAs with the specificity and sensitivity of TaqMan Assay chemistry. A simple, two-step protocol requires only reverse transcription with an miRNA-specific primer, followed by real-time PCR with TaqMan probes.

TaqMan MicroRNA Assays are:

  • Highly specific—quantitate only mature miRNAs, not precursors
  • Sensitive—conserve limited samples, requires only 1–10 nanograms of total RNA or equivalent
  • Fast, simple and scalable—two-step quantitative RT-PCR assay provides high-quality results in less than three hours

Our TaqMan MicroRNA Assay catalogue is kept up to date with the Sanger database to provide comprehensive coverage with the most current annotation.

Availability and sizes

Made-to-Order assays are available in four different sizes and usually ship within two weeks. Inventoried assays are prepackaged in Small size and are ready to ship. Inventoried assays may be purchased in larger sizes by ordering them as Made to Order.

Species

TaqMan MicroRNA Assays are available for a range of species including human, mouse, rat, Drosophila, C. elegans, and Arabidopsis. We will continue to increase the number of miRNA assays for these species in alignment with the Sanger miRBase Registry.

Assay Components

TaqMan MicroRNA Assays are delivered in two tubes: one containing the RT primer, the other containing the specific pre-formulated TaqMan Assay (TaqMan probe and PCR primer set).

Pre-formulated assay

  • Forward primer: 1500 nM
  • Reverse primer: 700 nM
  • FAM-MGB probe: 200 nM
  • RT sequences: 50 nM

TaqMan miRNA Assay selection guide

TaqMan MicroRNA Assays

  • Description: TaqMan MicroRNA Assays employ a novel target-specific stem–loop primer during cDNA synthesis to produce a template for real-time PCR
  • RT chemistry: miRNA-specific RT
  • Throughput: Best for 1–10 targets
  • Coverage: 205 species available, coverage for miRBase v.21

TaqMan Advanced miRNA Assays

  • Description: TaqMan Advanced miRNA Assays employ a universal RT step for a streamlined workflow, and a universal miR-Amp step to enable highly sensitive detection by real-time PCR
  • RT chemistry: Universal RT
  • Throughput: Best for >10 targets
  • Coverage: All human, mouse, and rat miRNAs; coverage for miRBase v.24
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionXS (25 RT/75 PCR reactions), Made to Order
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
FormatTube
No. of Reactions75 Reactions
Product LineTaqMan
Product TypeMicrorna Assay
QuantityXS (25 RT/75 PCR reactions), Made to Order
Shipping ConditionRoom Temperature
Sufficient For75 Reactions
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
• 1 tube containing RT primer at concentration of 5X (XS and S sizes), 20X (M size), or 60X (L size)
• 1 tube containing a 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15 to -25°C.

Frequently asked questions (FAQs)

What are the performance specifications of the TaqMan miRNA assays?

The TaqMan miRNA asays are guaranteed to meet the following:
Dynamic Range: > 6 logs10 with > 0.97 linearity (R2 value)
Specificity: Majority of assays have < 5% cross reactivity with closely related sequences. NTC background: Ct > 38.0
Lot-to-Lot Reproducibility: Difference between Ct's < 0.6 Ct when different lots of an assay are run with the same sample and master mix from the same lot on the sample plate
Amplification Efficiency: ranging from 90-110% across 5 logs10

Please see the document “TaqMan Assays QPCR Guarantee Program” for more details.

What data analysis tools do you recommend?

If you are using an Applied Biosystems instrument, we recommend using Expression Suite to analyze data from the TaqMan MicroRNA Array Cards. After importing the .sds or .eds files, you can perform all the QC and data analysis within the tool. For more details on how to use Expression Suite software, please see this video series: https://learn.thermofisher.com/courses/view/id/325

Do you have data to back up your claim that your TaqMan MicroRNA Assays can accurately distinguish miRNA targets that differ by a single base? Have you tested each TaqMan MicroRNA Assay that is designed to one of two or more closely-related target sequences?

It is well understood within the miRNA community that designing assays for miRNAs is challenging due to their short length (<22 bases) and closely related sequences. Although we have not tested cross reactivity of every closely related species, we have demonstrated that we can achieve <5% cross reactivity between a single nucleotide mismatch. Specificity of an assay depends on the number of mismatches to its closest homologue, the location of the mismatch, and the surrounding bases, making cross reactivity difficult to predict. As a general rule, the most difficult miRNA targets to discriminate are those with minimal mismatches localized to the 5' region of the sequence, and it is close to impossible to design an assay that discriminates between a single mismatch at the 5' most base. In addition, the assays in our catalog have been designed to provide a balance between specificity and sensitivity: an assay may be very specific but lack the needed sensitivity, or vice versa. To achieve this balance, and to ensure the highest sensitivity and to reduce false positives, TaqMan MicroRNA Assays must have an NTC background Ct > 38.0 and display good linearity across at least 3 logs10 (ideal R2 > 0.98).

What about isomiRs? Will Thermo Fisher Scientific assays give me good quantification if they only detect one isomer and not all of them?

Deep sequencing analysis of mature miRNAs revealed that many miRNAs have either an addition or deletion of 1-3 bases at the 3' and less frequently at the 5' terminal end. These are often referred to as isomiRs. The sequence deposited in miRBase is the canonical sequence derived by aligning sequences from current deep sequencing data. Thus far, there has been no biological relevance attached to these different forms since they exclusively occur outside the seed sequence. For that reason, the changes detected in the expression level of one isomer are proportional to changes within the entire pool. As a result, there may be a shift in raw Ct value using assays targeting two separate isomiRs. However, the relative expression ddCt has been demonstrated to be roughly the same. It should be noted that, although TaqMan MicroRNA Assays are designed to be sequence specific, they will detect a small spectrum of isomiRs. Depending on the number and composition at the 3' end, an assay may detect the +1 and +2 isomiRs but not the -1 or -2 forms.

What I can do to minimize variability when using assays?

Use multiple replicates and consider, when possible, using an overall study control that is used in every assay to monitor potential day-to-day/run-to-run/across study variability.