Pair your qPCR Master Mix with ready to use TaqMan Gene Expression, Genotyping Assay or Custom Probes for multiplexing. Use our helpful TaqMan Search Wizard to find the Assays for your target of interest. Learn more
TaqMan™ Fast Virus 1-Step Master Mix for qPCR
TaqMan™ Fast Virus 1-Step Master Mix for qPCR
TaqMan™ Fast Virus 1-Step Master Mix for qPCR
Applied Biosystems™

TaqMan™ Fast Virus 1-Step Master Mix for qPCR

TaqMan Fast Virus 1-Step Master Mix is designed for fast, highly sensitive real-time RT-PCR, even in the presence of challenging PCR inhibitors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
44444361 x 10 mL
44444345 x 1 mL
44444321 x 1 mL
Catalog number 4444436
Price (USD)
3,973.05
Special offer
Online exclusive
Ends: 11-Dec-2026
4,905.00
Save 931.95
Each
Quantity:
1 x 10 mL
Price (USD)
3,973.05
Special offer
Online exclusive
Ends: 11-Dec-2026
4,905.00
Save 931.95
Each

The features of the kit have been selected to enhance virus detection on commonly used sample types. A single-tube format allows for uniform handling and processing for both RNA and DNA viruses.

TaqMan Fast Virus 1-Step Master Mix is designed for fast, highly sensitive real-time RT-PCR even in the presence of challenging PCR inhibitors. The 4X formulation provides enhanced detection of both RNA and DNA viruses and is ideal for multiplex gene expression studies, even with low target input. The TaqMan Fast Virus 1-Step Master Mix provides a single-tube format for uniform handling and processing, while delivering qPCR cycling in less than 30 minutes, helpful when working in high throughput workflows.

Key product features:

  • High sensitivity—4X master mix to amplify both RNA and DNA with improved sensitivity
  • Consistent results—formulated to handle common RT-PCR inhibitors found in blood, stool, and other difficult samples
  • Simplified application— one-tube, one-step master mix, offering a single run profile with RNA and DNA, which allows for easy mix-and-match of targets on a plate
  • Target flexibility—works in singleplex, multiplex, and with exogenous or endogenous internal controls
  • High efficiency—increased qRT-PCR speed on fast and on standard instruments

Two formulations

The master mix is available in two formulations:

  • TaqMan Fast Virus 1-Step Master Mix containing ROX dye as a passive reference is recommended for singleplex (1 probe) and up to triplex (3 probes) reactions
  • TaqMan Fast Virus 1-Step Multiplex Master Mix (No ROX) (Cat. No. 5555532), which does not contain a passive reference, is recommended for use with assays utilizing the JUN (or similar omission wavelength) dye or for higher order multiplexing (>3 targets)

High sensitivity

Many common research samples with viral nucleic acids have very low levels of target. We have optimized the master mix for high-sensitivity detection of viral targets. The higher-concentration master mix allows you to set up smaller reactions, which means that you can perform fast cycling protocols and obtain at least the same sensitivity as is expected from standard-cycling qPCR. Alternatively, larger sample input amounts can be added to standard reaction volumes for more accurate quantification of low-titer samples (figure below).

Consistent results in the presence of inhibitors

Research samples commonly assayed for viruses include blood, dirt, and tissues. Buffer components and proprietary additives in the TaqMan Fast Virus 1-Step Master Mix have been optimized to handle RT-PCR inhibitors (figure below) to help ensure consistent performance even with these difficult samples, so that you can be more confident in your results.

Flexibility in targets

It is common for labs to test for both RNA and DNA viruses in a variety of samples. To simplify your experiments, a single TaqMan Fast Virus 1-Step Master Mix protocol has been developed to assay both types of nucleic acid, so you can perform RNA and DNA virus queries next to each other on the same plate using the same handling steps (figure below). Additionally, because virus research often includes multiplexed primers and probes and internal reaction controls, we have optimized the master mix to work with multiple target amplicons (figure below).

Fast results

The TaqMan Fast Virus 1-Step Master Mix speeds your time to results and maximizes the use of your real-time PCR instruments with a fast protocol. The high-concentration formulation allows for more target nucleic acid sample to be added in the smaller reaction volumes required to run fast protocols. This enables you to maintain sensitivity with low-titer research samples while improving speed and throughput (figure below).

TaqMan Fast Virus 1-Step Master Mix provides a reliable and efficient reagent for real-time RT-PCR of virus samples that does not sacrifice accuracy. Its robust performance in the presence of common RT-PCR inhibitors and its convenient and flexible reaction setup allows you to have more confidence in your results.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT System, QuantStudio 12k Flex, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio 7 Flex, StepOne, StepOnePlus, ViiA 7 System, Applied Biosystems AmpQuest 96
FormatTube
Hot StartBuilt-In Hot Start
Inhibitor Tolerance (Simple)High
Multiplex CapabilityUp to 3-plex
No. of Reactions2000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq Fast DNA Polymerase
Product LineTaqMan
Product Type1-Step Master Mix
Quantity1 x 10 mL
Reverse TranscriptaseM-MLV
Sample TypeTotal RNA, Viral DNA, Viral RNA, mRNA
Shipping ConditionDry Ice
Sufficient For2000 Reactions
Target SpecificityRNA only, Both RNA and DNA
ThermostabilityNot Applicable
Detection MethodPrimer-probe
For Use With (Application)Microbial Detection, Gene Expression, DNA Quantification, Presence/Absence
FormLiquid
PCR Method1-step RT-qPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
Contains 1 x 10 mL tube of 4X RT-PCR master mix sufficient for 2000 x 20 μL reactions. Components:
• AmpliTaq™ Fast DNA Polymerase, UP
• Thermostable MMLV Reverse Transcriptase
• dNTPs
• RNase Inhibitor (RNaseOUT™)
• ROX™ dye (passive reference)

Store at -15°C to -25°C in the dark.

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.