Product Image
Applied Biosystems™

Megaplex™ RT Primers, Human Pool Set v3.0

Designed to streamline experimental workflows, Megaplex™ RT Primers provide a single reaction solution when preparing cDNA for real-time PCR analysisRead more
Have Questions?
Catalog NumberQuantity
444474550 reactions
Catalog number 4444745
Price (USD)
1,956.00
Each
Quantity:
50 reactions
Price (USD)
1,956.00
Each
Designed to streamline experimental workflows, Megaplex™ RT Primers provide a single reaction solution when preparing cDNA for real-time PCR analysis on a TaqMan® MicroRNA Array. The Human Pool Set v3.0 contains two Megaplex™ RT Primer Pools, Pools A v2.1 and B v3.0. Collectively the pools cover 754 unique microRNAs. Additionally, each pool contains the same 4 controls. The microRNA targets represented in Human Pool A tend be better characterized, more broadly expressed and⁄or expressed at a higher level when compared to the Human Pool B while the microRNA targets represented in Human Pool B tend to be less well characterized, more narrowly expressed and⁄or expressed at a lower level compared to the Human Pool A. The Human Pool Set v3.0 is intended for use with the TaqMan® Array Human MicroRNA Card Set v3.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions50 Reactions
PrimerRT Primers
Product LineMegaplex
Product TypeRT Primer
Quantity50 reactions
SpeciesHuman
Sufficient For50 Reactions
For Use With (Application)miRNA analysis
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -20°C.

Frequently asked questions (FAQs)

Can Ct's greater than a cut-off be considered valid results?

Yes they can. However, it is important to recognize the true linearity and detection limits of your assay: Ct values above the cut-off can indicate non-specific amplification, unless your NTC is a true- no-target control, and you have run a statistically significant number of replicates. Any results with Ct above the recommended cut-off need to be validated with individual assays on plates.

What is a good Ct cut-off for the TaqMan MicroRNA Array Cards and TaqMan Advanced miRNA Array Cards? In other words, beyond what Ct should I not trust the data?

The typical Ct cut-off on TaqMan Array Cards is 32, which is equivalent to Ct 35 on a plate (10 µl reaction). Previous studies show that if you use pre-amplification, a Ct cut-off of 29 or 30 can be used to reduce numbers of false positives (see Technical Note Optimized protocols for human or rodent microRNA profiling with precious samples). To ensure that you have selected a correct cut-off, you should run replicates of the same sample and use Ct cut-off before you see an increase in the Standard Deviation.

Why does the negative control well show amplification when doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards?

Most likely the reagents or the cDNA template are contaminated. Please follow established PCR laboratory best practices.

Why do I have poor reproducibility across technical replicates when doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards?

Most likely the reagents were not adequately mixed. Ensure that all samples and reagents are mixed well.

When doing microRNA analysis using Megaplex Primer Pools and TaqMan Array Cards, why are the Ct values for the endogenous controls highly variable across the sample set?

Highly variable values for endogenous controls is most likely due to biological variation. We recommend that you use an alternative endogenous control, such as a non-variable miRNA.