Product Image
Applied Biosystems™

Megaplex™ Primer Pools, Human Pools Set v3.0

Megaplex™ Primer Pools, Human Pools Set v3.0 is available under a single part number containing Megaplex™ RT Primers Human PoolRead more
Have Questions?
Catalog NumberQuantity
444475050 reactions
Catalog number 4444750
Price (USD)
2,055.00
Each
Quantity:
50 reactions
Price (USD)
2,055.00
Each
Megaplex™ Primer Pools, Human Pools Set v3.0 is available under a single part number containing Megaplex™ RT Primers Human Pool A v2.1, Megaplex™ RT Primers Human Pool B v3.0, Megaplex™ PreAmp Primers, Human Pool A v2.1 and Megaplex™ PreAmp Primers, Human Pool B v3.0. Megaplex™ Primer Pools, Human Pools Set v3.0 contains RT primers for 754 unique microRNAs plus 4 controls. For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The Megaplex™ Primer Pools, Human Pools Set v3.0 is intended for use with the TaqMan® Array Human MicroRNA Card Set v3.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan® MicroRNA Assays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions50 Reactions
PrimerRT Primers, PreAmp Primers
Product LineMegaplex
Product TypePrimer Pool
Quantity50 reactions
SpeciesHuman
Sufficient For50 Reactions
For Use With (Application)miRNA analysis
GC-Rich PCR PerformanceLow
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Store at -20°C

Frequently asked questions (FAQs)

Can Ct's greater than a cut-off be considered valid results?

Yes they can. However, it is important to recognize the true linearity and detection limits of your assay: Ct values above the cut-off can indicate non-specific amplification, unless your NTC is a true- no-target control, and you have run a statistically significant number of replicates. Any results with Ct above the recommended cut-off need to be validated with individual assays on plates.

What is a good Ct cut-off for the TaqMan MicroRNA Array Cards and TaqMan Advanced miRNA Array Cards? In other words, beyond what Ct should I not trust the data?

The typical Ct cut-off on TaqMan Array Cards is 32, which is equivalent to Ct 35 on a plate (10 µl reaction). Previous studies show that if you use pre-amplification, a Ct cut-off of 29 or 30 can be used to reduce numbers of false positives (see Technical Note Optimized protocols for human or rodent microRNA profiling with precious samples). To ensure that you have selected a correct cut-off, you should run replicates of the same sample and use Ct cut-off before you see an increase in the Standard Deviation.

Why are the Ct values for my endogenous controls are highly variable across the sample set?

Highly variable values for endogenous controls is most likely due to biological variation. Use an alternative endogenous control such as a non-variable miRNA.

Why is the Ct value for the No Template Control (NTC) <35 for some assays?

Please review the following possible causes and solutions:
- There are non-specific interactions between primers. For NTC information on a specific assay, refer to the Megaplex Assay Performance File.
- The cDNA template is contaminated. Please follow established PCR good laboratory practices.
- The preamplification product was not diluted properly before real-time PCR. Follow the recommendations in the protocol. Please see "Dilute the preamplification reaction" on page 18.

Why did I not detect any miRNA?

It is possible that the threshold is set too high to detect miRNA in samples with low expression. Identify an appropriate NOAMP flag threshold if the relative threshold algorithm (Crt) is used. Alternatively, identify an appropriate threshold if Ct is used.