TaqMan™ Gene Expression Assays
Applied Biosystems™

TaqMan™ Gene Expression Assays

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityLabel or Dye
444889275 Reactions, Made to OrderFAM
4331182250 Reactions, InventoriedFAM
43513682900 Reactions, Made to OrderFAM
4351370750 Reactions, Made to OrderFAM
4351372360 Reactions, Made to OrderFAM
445332075 Reactions, InventoriedFAM
A80710S (360 Reactions), Made to OrderFAM
A80711M (750 Reactions), Made to OrderFAM
A80712L (2900 Reactions), Made to OrderFAM
A80716S (360 Reactions), Made to OrderABY
A80717M (750 Reactions), Made to OrderABY
A80718L (2900 Reactions), Made to OrderABY
A80719S (360 Reactions), Made to OrderJUN
A80720M (750 Reactions), Made to OrderJUN
A80721L (2900 Reactions), Made to OrderJUN
A80722S (360 Reactions), Made to OrderCy5
A80723M (750 Reactions), Made to OrderCy5
A80724L (2900 Reactions), Made to OrderCy5
Catalog number 4448892
Price (USD)
108.65
Online exclusive
116.00
Save 7.35
Each
Quantity:
75 Reactions, Made to Order
Label or Dye:
FAM

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM, VIC, ABY, JUN, and Cyanine 5) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Features of TaqMan Gene Expression assays include:

  • Easy to use—just add cDNA and master mix and run—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Extensive content—Over 1.8 million predesigned assays available for over 25 different species
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Fast delivery—your assays shipped in as little as 1–2 days (see below)
  • Multiplexing ready—TaqMan assays are now available with 5 different dye labels to support simultaneous detection of up to 5 targets in a single qPCR reaction. FAM, ABY, JUN, and Cyanine 5 assays are available with balanced primer/probe concentrations (Primer-Limited, 350 nM) to support consistent multiplex performance. VIC Primer-Limited assays (150 nM) are available with SKU 4448484 (S), 4448485 (M), 4448486 (L).

Approximate ship time

  • Inventoried: 1–2 days in North America, 3–5 days in Europe*
  • Made to order: 4–6 days in North America, 6–10 days in Europe (FAM, VIC); 4-10 days in North America, 6-14 days in Europe (ABY, JUN, Cyanine 5)

TaqMan Gene Expression assays are the gold standard in qPCR and dPCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Pre-formulated assay

  • 2 unlabeled PCR primers (Standard: 900 nM each final 1X concentration; Primer-Limited: 350 nM each final 1X concentration)
  • 1 dye-labeled (FAM, VIC, ABY, JUN or Cyanine 5) TaqMan MGB probe (250 nM final 1X concentration)

All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays Guarantee.**

*Our most popular assays are stocked in North America and Europe for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556) or TaqPath DuraPlex 1-Step RT-qPCR Master Mix (Cat. No. A58666)

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Fidelity (vs. Taq)60 X
For Use With (Equipment)7500 System, 7500 Fast System, 7900HT System, StepOne™, StepOnePlus™, ViiA™ 7 System, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio™ 12k Flex, QuantStudio™ Absolute Q Digital PCR System
FormatTube
No. of Reactions75 Reactions, Made to Order
Product LineTaqMan
Product TypeGene Expression Assay
Quantity75 Reactions, Made to Order
Shipping ConditionRoom Temperature
Concentration20X
Detection MethodPrimer-probe
GC-Rich PCR PerformanceHigh
Label or DyeFAM
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).

Store at -15°C to -25°C.

Frequently asked questions (FAQs)

Can I multiplex my assays if my amplicons are not all the same length?

Yes, it is possible. The amplicons do not have to be the same length in order to duplex the assays. You need to use different reporter dyes on the probes in multiplex - they should have a substantial difference in maximum emission wavelength to be effective. A classic example is to use FAM and VIC for the 2 probes. Depending on the expression of the targets, you may also need to limit the primer concentration for one of the assays. This is common when duplexing a target and endogenous control (the control probe is VIC labeled and the primer concentration is decreased so the target assay can properly compete for use of enzyme and dNTPs).

I want to order a TaqMan Gene Expression Assay that you have discontinued. Is there a way do that?

Yes, you can order it as a custom assay. Since the assay is discontinued, it can not be ordered on-line. You will have to call or email the order to genomics@appliedbiosystems.com, and include the assay ID. The assay will be synthesized once the order is received. It will be designed and priced as a Custom TaqMan Gene Expression Assay so you will need to choose the scale.

Can I use TaqMan Gene Expression Assays for a gene copy number study?

TaqMan Gene Expression Assays with “_s1” as assay ID suffix are the assays whose primers and probes are designed within a single exon, and therefore, will detect genomic DNA. However, the application of these assays in gene copy number studies has not been validated. Other gene expression assays will not detect gDNA, let alone the region of variance.

I am working on both mouse and human samples for gene expression studies. Do you have one endogenous control that I can use with both species?

The 18S rRNA is highly conserved among eukaryotes. We have several configurations of assays for this gene: P/N 4331182, Assay ID: Hs99999901_s1; PN 4319413E, 4310893E, 4333760T, 4333760F, 4308329. You can use any of these for both human and mouse samples, as well as samples from any other eukaryotic species.

Can I check the end product of a TaqMan Gene Expression Assay by running an agarose gel?

Yes, you can check the end product on an agarose gel. If you use the TaqMan Universal PCR Master Mix (including AmpErase UNG, Cat. No. 4304437), we recommend running the gel immediately after the PCR, since the trace amount of AmpErase UNG may digest the end product that contains double-stranded DNA with dUTP incorporated. The average length of amplicon ranges from 50-150 bp. The exact amplicon length is indicated on the detailed page of each assay on our website. You can get to that page by clicking on the Assay ID; the amplicon information is listed in the section "Assay Details".