Immuno Breakable Modules Clear, Nunc; U8 LockWell; PolySorp; 320μL - FAQs
Immuno Breakable Modules Clear, Nunc; U8 LockWell; PolySorp; 320μL - FAQs
View additional product information for Immuno Breakable Modules Clear - FAQs (473539, 473768, 446469, 446639, 446490, 446477, 446442, 95029390)
1 product FAQs found
Are your Thermo Scientific 8-Well Strip Caps for Immuno Standard Modules compatible with LockWell modules?
Due to the locking feature design of the LockWell modules, the 8-Well Strip Caps for Immuno Standard Modules are not compatible with, and will not seal, the LockWell Modules.