TaqMan™ Mutation Detection Assay with castPCR™ Technology
Applied Biosystems™

TaqMan™ Mutation Detection Assay with castPCR™ Technology

Powered by competitive allele-specific TaqMan™ PCR (castPCR™ Technology) to detect and measure somatic mutations in genes associated with cancer research.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
4465807CNGene Reference (75 Reactions), Inventoried
4465804CNMutant Allele (75 Reactions), Inventoried
Catalog number 4465807CN
Price (USD)
-
Quantity:
Gene Reference (75 Reactions), Inventoried

Applied Biosystems TaqMan Mutation Detection Assays are powered by competitive allele-specific TaqMan castPCR technology to detect and measure somatic mutations in genes associated with cancer research. castPCR technology is highly specific and sensitive, and can detect rare amounts of mutated DNA in a sample that contains large amounts of normal, wild-type DNA. These assays are compatible with different sample types, such as cell lines, formalin-fixed paraffin-embedded (FFPE) tissue samples, and fresh-frozen tissue samples.

castPCR technology

Competitive, allele-specific TaqMan PCR utilizes an allele-specific primer for mutant allele detection that competes with an MGB blocker oligonucleotide to suppress the wild-type background.

Somatic mutation detection

By combining the power of TaqMan and castPCR technologies, TaqMan Mutation Detection Assays provide:

  • Higher specificity—designed to block the wild-type and amplify only the mutant
  • Higher sensitivity—designed to detect somatic mutations down to 1 cancer cell in 1,000 normal cells
  • Faster workflow—3 hours from sample to result

With 819 assays for 47 known cancer research genes available today like KRAS, BRAF, KIT, JAK2, the TaqMan Mutation Detection Assays provide an innovative tool for your cancer research.

All TaqMan Mutation Detection Assays have undergone extensive testing to ensure verify high sensitivity and specificity.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatTube
No. of Reactions75 Reactions
Product LineTaqMan
Product TypeMutation Detection Assay
QuantityGene Reference (75 Reactions), Inventoried
Shipping ConditionRoom Temperature
Volume150 μL
Concentration10X
Detection MethodPrimer-probe
FormFrozen
PCR MethodqPCR
Reaction SpeedFast
Unit SizeEach
Contents & Storage
1 tube containing a 10X mix of unlabeled PCR primers and a TaqMan MGB probe

Store at -15 to -25°C.