Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SRC Antibodies

          Invitrogen

          Phospho-SRC (Tyr419) Polyclonal Antibody

          54 Published Figures
          157 References
          View all (73) SRC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-SRC (Tyr419) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (54)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 56

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-SRC (Tyr419) Antibody (44-660G) in IHC (P)

          Immunohistochemistry analysis of SRC showing staining in the cytoplasm and membrane of paraffin-embedded human colon carcinoma (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for 15 min at room te... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-SRC (Tyr419) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-SRC (Tyr419) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-SRC (Tyr419) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-SRC (Tyr419) Antibody in Western Blot, Immunohistochemistry (Paraffin) (WB, IHC (P))
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Immunohistochemistry (IHC)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)
          Phospho-SRC (Tyr419) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-SRC (Tyr419) Polyclonal Antibody

          Product Details

          44-660G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 96 publications 96 publications

          Immunohistochemistry (IHC)

          -
          View 20 publications 20 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 19 publications 19 publications

          ELISA (ELISA)

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 10 publications 10 publications

          Miscellaneous PubMed (Misc)

          -
          View 9 publications 9 publications
          Product Specifications

          Species Reactivity

          Chicken, Human, Mouse

          Published species

          Bovine, Dog, Fruit fly, Human, Mouse, Pig, Rat, Tag, Xenopus, Yeast, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human Src that contains tyrosine 419. The sequence is conserved in mouse, rat, chicken and frog.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533714

          Target Information

          SRC is a cytoplasmic tyrosine kinase and proto-oncogene involved in the regulation of embryonic development and cell growth. SRC is involved in the regulation of growth and differentiation of eukaryotic cells. SRC associates with the cytoplasmic region of various cell surface receptors to facilitate signal transduction, and is also involved in cytoskeletal organization, cell cycle regulation, and apoptosis. In humans, the gene encoding SRC is present on chromosome 20. Activity of SRC is regulated by two tyrosine residues (Tyr 416 and Tyr527), and phosphorylation of these two sites have opposing effects. Phosphorylation of Tyr416 in the activation loop of the kinase domain upregulates enzyme activity while the phosphorylation of Tyr527 decreases enzyme activity. Mutations in the SRC gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same SRC protein have been found thus far.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: c-src protein, C terminus; EC 2.7.10.2; neuronal proto-oncogene tyrosine-protein kinase Src; non-tyrosine protein kinase; ORF; OTTHUMP00000174476; OTTHUMP00000174477; OTTHUMP00000174478; p60-Src; pp60c-scr; pp60c-src; pp60src; proto-oncogene c-Src; protooncogene SRC, Rous sarcoma; Rous sarcoma oncogene; RP5-823N20.1; src downstream region; tyrosine kinase pp60c-src; tyrosine-protein kinase SRC-1; tyrosine-specific protein kinase; Unknown; unnamed protein product; v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog; v-src sarcoma viral oncogene

          View more View less

          Gene Aliases: ASV; c-SRC; p60-Src; PP60C-SCR; pp60c-src; SDR; SRC1; THC6

          View more View less

          UniProt ID: (Chicken) P00523, (Human) P12931, (Mouse) P05480

          View more View less

          Entrez Gene ID: (Chicken) 396442, (Human) 6714, (Mouse) 20779

          View more View less

          Function(s)
          protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity receptor binding protein binding ATP binding protein phosphatase binding heme binding growth factor receptor binding connexin binding protein kinase activity integrin binding phospholipase activator activity kinase activity enzyme binding protein domain specific binding protein tyrosine kinase activator activity receptor activator activity ionotropic glutamate receptor binding SH2 domain binding phospholipase binding ion channel binding cadherin binding ephrin receptor binding ATPase binding phosphoprotein binding BMP receptor binding scaffold protein binding protein kinase C binding insulin receptor binding protein kinase binding estrogen receptor binding macromolecular complex binding cell adhesion molecule binding transmembrane signal receptor scaffold/adaptor protein
          Process(es)
          cell cycle cell adhesion transmembrane receptor protein tyrosine kinase signaling pathway epidermal growth factor receptor signaling pathway cell differentiation osteoclast development peptidyl-tyrosine autophosphorylation regulation of cell proliferation innate immune response bone resorption progesterone receptor signaling pathway regulation of cell cycle negative regulation of extrinsic apoptotic signaling pathway negative regulation of intrinsic apoptotic signaling pathway negative regulation of transcription from RNA polymerase II promoter stimulatory C-type lectin receptor signaling pathway negative regulation of inflammatory response to antigenic stimulus signal transduction signal complex assembly transforming growth factor beta receptor signaling pathway integrin-mediated signaling pathway lactation regulation of epithelial cell migration positive regulation of epithelial cell migration macroautophagy peptidyl-tyrosine phosphorylation regulation of cell-cell adhesion platelet activation osteoclast differentiation T cell costimulation negative regulation of protein complex assembly protein destabilization negative regulation of telomere maintenance positive regulation of integrin activation regulation of toll-like receptor 3 signaling pathway cellular response to reactive oxygen species positive regulation of Rac protein signal transduction positive regulation of dephosphorylation negative regulation of hippo signaling intracellular signal transduction Fc-gamma receptor signaling pathway involved in phagocytosis ERBB2 signaling pathway angiotensin-activated signaling pathway vasodilation negative regulation of apoptotic process regulation of vascular permeability stress fiber assembly regulation of bone resorption positive regulation of Notch signaling pathway positive regulation of glycolytic process viral entry into host cell vascular endothelial growth factor receptor signaling pathway ephrin receptor signaling pathway focal adhesion assembly leukocyte migration positive regulation of small GTPase mediated signal transduction negative regulation of focal adhesion assembly positive regulation of protein kinase B signaling negative regulation of mitochondrial depolarization intestinal epithelial cell development interleukin-6-mediated signaling pathway positive regulation of ERK1 and ERK2 cascade response to interleukin-1 cellular response to peptide hormone stimulus cellular response to progesterone stimulus regulation of heart rate by cardiac conduction negative regulation of neutrophil activation positive regulation of TORC1 signaling positive regulation of lamellipodium morphogenesis regulation of early endosome to late endosome transport negative regulation of anoikis regulation of caveolin-mediated endocytosis primary ovarian follicle growth positive regulation of cytokine production protein phosphorylation gamete generation positive regulation of cell proliferation positive regulation of gene expression negative regulation of gene expression positive regulation of glucose metabolic process positive regulation of protein processing positive regulation of smooth muscle cell migration cell migration protein deubiquitination positive regulation of cell migration forebrain development cellular response to nutrient levels regulation of intracellular estrogen receptor signaling pathway toll-like receptor 3 signaling pathway substrate adhesion-dependent cell spreading cellular response to platelet-derived growth factor stimulus TORC1 signaling odontogenesis positive regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of MAP kinase activity transcytosis positive regulation of bone resorption positive regulation of cell adhesion positive regulation of cell size positive regulation of Ras protein signal transduction positive regulation of insulin receptor signaling pathway neurotrophin TRK receptor signaling pathway oogenesis positive regulation of peptidyl-tyrosine phosphorylation positive regulation of protein transport uterus development branching involved in mammary gland duct morphogenesis regulation of cell projection assembly cellular response to hydrogen peroxide cellular response to fluid shear stress cellular response to transforming growth factor beta stimulus positive regulation of podosome assembly DNA biosynthetic process positive regulation of canonical Wnt signaling pathway positive regulation of wound healing positive regulation of protein localization to nucleus positive regulation of intracellular signal transduction positive regulation of cellular protein localization negative regulation of TORC1 signaling cellular response to angiotensin positive regulation of vascular smooth muscle cell proliferation positive regulation of neural precursor cell proliferation positive regulation of male germ cell proliferation positive regulation of ovarian follicle development positive regulation of platelet-derived growth factor receptor-beta signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch