TaqMan™ GMO Screening Kit - FAQs
TaqMan™ GMO Screening Kit - FAQs
View additional product information for TaqMan™ GMO Screening Kit - FAQs (4466334)
1 product FAQs found
What is the TaqMan GMO Screening kit designed for?
The TaqMan GMO Screening kit is designed to perform four reactions for each sample analysed (P35S, TNOS, P34S-FMV, and plant).