MicroSEQ™ ID Microbial Identification Software v3.0 (3 licenses) - FAQs
MicroSEQ™ ID Microbial Identification Software v3.0 (3 licenses) - FAQs
View additional product information for MicroSEQ™ ID Microbial Identification Software v3.0 (3 licenses) - FAQs (4467864)
1 product FAQs found
Which 3500 Data Collection software version is compatible with MicroSEQ ID Microbial Identification Software v3.0?
MicroSEQ ID Microbial Identification Software v3.0 is only compatible with 3500 Data Collection software version 1.1.