Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • JNK1/JNK2 Antibodies

          Invitrogen

          Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody

          Advanced Verification
          53 Published Figures
          69 References
          View all (12) JNK1/JNK2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (3)
          • Published Figures (53)
          • Advanced Verification (1)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 57

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody (44-682G) in ICC/IF

          Immunofluorescent analysis of JNK1/2 (pT183/pT185) was done on 70% confluent log phase A549 cells treated with Anisomycin (25 µg/mL for 30 min). The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with JNK1/2 (pT183/pT185) Rabbit polyclonal Antibody (Product # 44-68... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunohistochemistry (IHC)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Immunohistochemistry (IHC)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody in Western Blot (WB)
          Phospho-JNK1/JNK2 (Thr183, Tyr185) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-JNK1/JNK2 (Thr183, Tyr185) Polyclonal Antibody

          Product Details

          44-682G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 57 publications 57 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:250
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Dog, Human, Insect, Mouse, Non-human primate, Pig, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human JNK1&2 that contains threonine 183 and tyrosine 185. This region is conserved among many species including mouse, rat, chicken, nematode, fruit fly, and in JNK3.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533720

          Product Specific Information

          This antibody is reactive to human and rat JNK1&2. Other species of JNK1&2 have not been tested, and JNK3 (found primarily in neuronal cell lines) has not been detected. The antibody has been negatively preadsorbed using a non-phosphopeptide corresponding to the site of phosphorylation to remove antibody that is reactive with non-phosphorylated JNK1&2. The final product is generated by affinity chromatography using a JNK1&2-derived peptide that is phosphorylated at threonine 183 and tyrosine 185. Positive controls used: HEK 293 +/- UV irradiation treatment; PC12 cells +/- sorbitol.

          Target Information

          C-Jun N-terminal Kinase (JNK) is also known as Mitogen-activated protein kinase (MAPK8). It belongs to the MAPK superfamily of stress-activated protein kinases. MAPKs are Serine-threonine protein kinases that are activated in response to a variety of extracellular stimuli, and mediate signal transduction from the cell surface to the nucleus. JNK is involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation and development. JNK pathways are activated by stress and inflammatory signals. JNK is expressed as ten different isoforms due to differential mRNA splicing. The predominant forms are JNK1 and JNK2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: C-jun kinase; c-Jun kinase 2; c-Jun N-terminal kinase 1; c-Jun N-terminal kinase 2; c-jun NH2-terminal kinase; JNK 55; JNK/SAPK alpha; JNK1 beta1 protein kinase; Jun kinase; JUN N-terminal kinase; MAP kinase 8; MAP kinase 9; MAPK; MAPK 8; MAPK 9; MAPK8; MAPK9; OTTHUMP00000019552; p54 gamma; p54-alpha; Phospho-JNK; PRKM; protein kinase JNK1; protein kinase mitogen-activated 8; protein kinase, mitogen-activated 9; SAPK 1; SAPK gamma; SAPK-alpha; stress activated protein kinase alpha II; stress-activated protein kinase 1; stress-activated protein kinase 1a; stress-activated protein kinase 1c; stress-activated protein kinase JNK1; stress-activated protein kinase JNK2; unnamed protein product

          View more View less

          Gene Aliases: JNK; JNK-46; JNK-55; JNK1; JNK1A2; JNK2; JNK21B1/2; JNK2A; JNK2ALPHA; JNK2B; JNK2BETA; p54a; p54aSAPK; PRKM8; PRKM9; SAPK; SAPK1; SAPK1a; SAPK1c

          View more View less

          UniProt ID: (Human) P45983, (Human) P45984, (Mouse) Q91Y86, (Rat) P49185, (Mouse) Q9WTU6, (Rat) P49186

          View more View less

          Entrez Gene ID: (Human) 5599, (Human) 5601, (Mouse) 26419, (Rat) 116554, (Mouse) 26420, (Rat) 50658

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity JUN kinase activity MAP kinase activity protein binding ATP binding enzyme binding protein phosphatase binding histone deacetylase regulator activity histone deacetylase binding protein serine kinase activity protein serine/threonine kinase binding protein serine/threonine/tyrosine kinase activity kinase activity kinesin binding mitogen-activated protein kinase kinase kinase binding
          Process(es)
          MAPK cascade protein phosphorylation response to oxidative stress JNK cascade JUN phosphorylation response to UV response to mechanical stimulus positive regulation of gene expression regulation of macroautophagy peptidyl-serine phosphorylation peptidyl-threonine phosphorylation positive regulation of cyclase activity positive regulation of cell killing negative regulation of protein binding regulation of protein localization cellular response to amino acid starvation cellular response to oxidative stress cellular response to reactive oxygen species Fc-epsilon receptor signaling pathway regulation of circadian rhythm positive regulation of apoptotic process negative regulation of apoptotic process positive regulation of protein metabolic process stress-activated MAPK cascade cellular response to lipopolysaccharide cellular response to mechanical stimulus cellular senescence energy homeostasis positive regulation of NLRP3 inflammasome complex assembly positive regulation of establishment of protein localization to mitochondrion release of cytochrome c from mitochondria positive regulation of protein phosphorylation positive regulation of macrophage derived foam cell differentiation neuron projection development positive regulation of prostaglandin biosynthetic process regulation of protein ubiquitination positive regulation of protein ubiquitination positive regulation of prostaglandin secretion positive regulation of proteasomal ubiquitin-dependent protein catabolic process positive regulation of chemokine production cellular response to UV regulation of mRNA stability positive regulation of cell differentiation positive regulation of transcription, DNA-templated regulation of JNK cascade response to cadmium ion modulation of synaptic transmission mRNA destabilization protein localization to tricellular tight junction positive regulation of podosome assembly inflammatory response to wounding extrinsic apoptotic signaling pathway positive regulation of cytokine production involved in inflammatory response positive regulation of apoptotic signaling pathway ossification neuron migration positive regulation of neuroblast proliferation double-strand break repair regulation of transcription, DNA-templated integrin-mediated signaling pathway neuromuscular junction development regulation of gene expression positive regulation of cardiac muscle cell apoptotic process positive regulation of cell migration positive regulation of microtubule polymerization positive regulation of glial cell apoptotic process ERBB signaling pathway response to hydrogen peroxide positive regulation of neuron apoptotic process NLRP3 inflammasome complex assembly regulation of neuron differentiation positive regulation of DNA replication regulation of centrosome cycle determination of dorsal identity dendrite morphogenesis SMAD protein signal transduction protein poly-ADP-ribosylation cellular response to hydrogen peroxide cellular response to nitric oxide cellular response to oxygen-glucose deprivation type B pancreatic cell apoptotic process neuronal stem cell population maintenance programmed necrotic cell death regulation of dense core granule transport protein localization to site of double-strand break positive regulation of determination of dorsal identity positive regulation of neuron migration response to xenobiotic stimulus response to toxic substance response to amine neuron development protein localization to mitochondrion cellular response to interleukin-1 cellular response to tumor necrosis factor cellular response to growth factor stimulus cellular hyperosmotic response cellular response to beta-amyloid response to tetrachloromethane response to nerve growth factor glial cell apoptotic process response to steroid hormone
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch