Product Image
Applied Biosystems™

QuantStudio™ 12K Flex OpenArray™ Genotyping Starter Kit

A variety of starter kits are available to introduce you to the various applications of the QuantStudio™12K Flex OpenArray™ sampleRead more
Have Questions?
Catalog NumberQuantity
44696051 kit
Catalog number 4469605
Price (USD)
3,525.00
Each
Quantity:
1 kit
Price (USD)
3,525.00
Each
A variety of starter kits are available to introduce you to the various applications of the QuantStudio™12K Flex OpenArray™ sample block. Each starter kit contains 2 arrays, plus sufficient reagents and accessories to perform two experiments. The Genotyping starter kit contains 2 genotyping training arrays. Note that the QuantStudio™12K Flex Software (included with the QuantStudio™ 12K Flex System) offers example experiment files for each application.

The accessories provided in the kit include: 6 lids and plugs, 6 immersion fluid syringes, 2 carriers, 1 box of Accufill™ System tips, and 384-well plates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)QuantStudio™ 12k Flex
Format384-Well Plate
Genotyping TargetSNPs (Known)
PolymeraseAmpliTaq Gold DNA Polymerase
Product LineOpenArray, QuantStudio
Product Type12K Flex Genotyping Starter Kit
Quantity1 kit
Sample TypeDNA (Genomic)
SoftwareQuantStudio™ 12K Flex Genotyper v1.2
SpeciesHuman
TechniqueSNP Genotyping (Real-Time PCR-Based)
Detection MethodPrimer-probe
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
• 2X TaqMan™ OpenArray™ Genotyping Master Mix
• TaqMan™ OpenArray™ Genotyping Training Plates (2 plates) containing TaqMan™ SNP Genotyping Assays
• QuantStudio™ 12K Flex OpenArray™ Accessories Starter Kit

Frequently asked questions (FAQs)

What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.

How do I download my Genotyping Applied Biosystems OpenArray SPF plate files?

You can download your SPF plate files from here (http://www.thermofisher.com/us/en/home/products-and-services/product-types/download-openarray-tpf-and-spf-plate-files.html).