Search
View additional product information for GeneMapper™ ID-X Software Hotfix_20110620 - FAQs (4470685)
1 product FAQs found
Unfortunately, there is no training or support for the GeneMapperID-X command line features.
We recommend referencing the GeneMapper 5 Installation and Administration User Guide, Appendix B for supported command line features and working with your informatics team for additional support.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.