Need help? Find instrument-specific tips, troubleshooting help, and technical resources within our new Real-Time PCR and Digital PCR Instruments Support Center.
Learn more about financing options and programs » Learn more
Product Image
Applied Biosystems™

QuantStudio™ 12K Flex Real-Time PCR System, OpenArray™ block, Accufill™ System

The QuantStudio™ 12K Flex system is a highly flexible, comprehensive real-time PCR platform that leverages the best of Applied BiosystemsRead more
Have Questions?
Catalog NumberQuantity
44710901 system
Catalog number 4471090
Price (USD)
-
Quantity:
1 system
The QuantStudio™ 12K Flex system is a highly flexible, comprehensive real-time PCR platform that leverages the best of Applied Biosystems real-time technology in a single instrument. The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray® plates, TaqMan® array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for your project whether it’s now or in the future, for maximum flexibility and consistency in your research. Note: this particular system includes an OpenArray block and AccuFill™ sample handling system. For a package that does not include an AccuFill system, please see Cat. No. 4472380.

Thermo Fisher Cloud
Thermo Fisher Cloud provides scientists with a secure place to store, analyze, and share data. Applied Biosystems qPCR Analysis Modules within the Thermo Fisher Cloud are web browser-based software tools that are flexible, fast and easy to use. These software tools are compatible with Mac or PC as well as most Applied Biosystems instrument file types.

The QuantStudio™ 12K Flex System’s intuitive software, easy touch screen setup, automation integration, effortless block exchange, and easy-to- use OpenArray® components are designed to let you get started right away and reduce hands on time. Plan for the future and save time, money, and space in your lab with the only all-in-one qPCR instrument enabling a broad range of applications with maximum throughput.

Flexibility & Scalability
• Includes five interchangeable blocks: The QuantStudio™ 12K Flex blocks accomodate 96-well, FAST 96-well, 384-well, TaqMan® array cards, and OpenArray® plates, for effortless scaling from 1 to 12,000 data points in a single 100 minute run. Increase throughput as you dive deeper into your research, using just one instrument. As you begin your project, start with 96-well format experiments and then scale up to a higher throughput to get the statistically significant answers you need. Additionally, you can easily validate and screen your targets of interest with a selected panel of genes using novel formats such as the TaqMan® array cards and OpenArray® plates.
• Compatible with a full range of TaqMan® assays: The QuantStudio™ 12K Flex System is optimized to enable clear, clean data for all TaqMan® assays that might be used across a broad range of applications, including gene expression, microRNA and small noncoding RNAs, SNP genotyping, protein analysis, protein thermal shift, high resolution melt, copy number variation, and pathogen detection.
• Delivers peace of mind: Leave early on a Friday and monitor runs from home, knowing that the system can safely store over 100 runs.

Horsepower
• Enhanced OptiFlex™ System: Building upon Applied Biosystems’ proprietary technology, the Enhanced OptiFlex™ System provides enhanced fluorescence detection for the accurate and sensitive data analysis necessary to collect 12,000 data points in a single 100 minute run.
• Unprecedented coverage and sample throughput: The OpenArray® format accelerates genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.
• Simple OpenArray® workflow: QuantStudio™ Open Array® plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the Accufill™ System. Easily load four arrays into the QuantStudio™ 12K Flex instrument to achieve over 12,000 data points. Integrated sample tracking software lets you easily track your samples throughout the course of your project.
• Tools to maximize your resources: New intuitive touch-screen interface and integrated calendar feature allows for scheduling time of use on the instrument. Additional analysis software such as ExpressionSuite v1.0 and Genotyper v1.1 is available to help you analyze hundreds of targets and samples and achieve the statistical power you need.

QuantStudio™ 12K Flex Software
• Intuitive interface and innovative design for novice and experienced users
• Convenient walk-away operation
• Improved data analysis across block types
• Fully compatible with high-throughput environments

Expandable Software Architecture
Optional add-ons are available to upgrade the existing software:
• 21 CFR Part 11 compliance: This optional SAE module assists with Security, Auditing, and Electronic signature of records for full data traceability.
• High Resolution Melt (HRM): Using the built-in MeltDoctor™ protocols and calibrations, you can spend less time optimizing the run and more time customizing your results. Define the number of variants you want to see by quickly changing analysis settings.
• ExpressionSuite software v1.0: our latest stand-alone gene expression software with enhanced analysis tools that can analyze over 200 384-well plates or 10 OpenArray® plates.

Learn more about available financing options and programs ›

Specifications
Dimensions51 cm (W) x 74 cm (H) x 67 cm (D)
Display TypeTouchscreen
Dynamic RangeUp to 7 Logs of Linear Dynamic Range (OpenArray™)
Electrical Requirements60 Hz
External ComputerComputer and Monitor
For Use With (Equipment)QuantStudio™
FormatOpenArray™ Plate
Other Format Options96-well Fast Block Upgrade Option, TaqMan™ Array Card Block Upgrade Option, 384-well Block Upgrade Option, 96-well Standard Block Upgrade Option
Product LineQuantStudio
Quantity1 system
SensitivityDown to 1 Copy
Voltage100 to 240 V
Weight69 kg (152 lb.)
Block FormatInterchangeable
Capacity4 OpenArray™ (3072 Through-Hole) Plates
Unit SizeEach
Contents & Storage
Without Power Cord

Frequently asked questions (FAQs)

How do I activate or renew my Vector NTI software license?

Please follow the instructions listed in this manual (https://licensing.appliedbiosystems.com/web/static/tools/manual.pdf).

For problems with license renewal, please send an email to instrumenthardwaresupport@thermofisher.com (for North America customers) or eurotech@thermofisher.com (for EMEA customers), with the computer ID and license key (AID-number) so you can be assisted.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

When using the QuantStudio 12K Flex Real-Time PCR System, why are there no CRT values upon export of my data?

When this happens, the analysis was set to use ‘Relative Threshold' but the CRT values do not show up in the exported data. (Note: To use ‘Relative Threshold', go to “Analysis Settings”, and under the default Ct Settings tab, choose ‘Relative Threshold' under “Algorithm Settings”.)

To export the data, go to ‘Export' and make sure the ‘Results' section is checked. You will notice that the column header will still say ‘Ct' even if you have CRT in the well table like above. If you export the data but notice that the numbers still correspond to the ‘Ct' values (and not the CRT values), then you will need to use the ‘Load Export Set' button. Just click ‘OK' to the pop-up window. and the changes will be applied. The next time you export, the values should reflect the CRT values.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What can I do if I get the Error - "Interrupted data file" in the QuantStudio12K Flex software?

Sometimes an error can occur when the file is downloaded to the computer. When this occurs you may see an abrupt break in the data. If you see this, you can repair the data by going to Analysis then Analyze from Raw Data. You should then see a fixed plot. Please note that you would need to have at least v1.2 of the QuantStudio 12K Flex software to use this feature. If you do not have this version, or if you do but the error persists, please send your data file to techsupport@thermofisher.com so that we can attempt to recover the data.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Is it possible to see the traces in an allelic discrimination experiment in the QuantStudio 12K Flex Real-Time PCR System software?

The ‘reveal traces' feature for genotyping runs allows you to trace the clusters throughout the PCR process. For some assays you may find better cluster discrimination at, say, cycle 40, as opposed to cycle 50. To use this feature, the amplification data needs to have been collected for the run.

- Go to 'Analysis Settings'.
- Under the default ‘Call Settings' tab, Choose 'Analyze Real-Time Run (?) Data'.
- Click ‘Apply Analysis Settings'.
- Under the Allelic Discrimination Plot, check the box next to ‘Reveal Traces'. You can then move the slide bar to see how the data change with the cycle number.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

How do I transfer a QuantStudio 12K Flex software license to another computer?

Please follow the directions in this user guide (https://licensing.appliedbiosystems.com/web/static/tools/manual.pdf) to manage your license (including transfer and renewing a license).

If you are unable to resolve the issue after following these steps, please contact us at instrumentservices@thermofisher.com for further assistance.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Citations & References (1613)

Citations & References
Abstract
Anticancer Activity of Ω-6 Fatty Acids through Increased 4-HNE in Breast Cancer Cells.
Authors:Bose C,Hindle A,Lee J,Kopel J,Tonk S,Palade PT,Singhal SS,Awasthi S,Singh SP
Journal:Cancers
PubMed ID:34944997
SIMPLE SUMMARY: Epidemiological evidence suggests that breast cancer risk is lowered by Ω-3 and increased by Ω-6 polyunsaturated fatty acids (PUFAs). Paradoxically, the Ω-6 PUFA metabolite 4-hydroxynonenal (4-HNE) inhibits cancer cell growth. This duality prompted us to study whether arachidonic acid (AA) would enhance doxorubicin (dox) cytotoxicity towards breast cancer ... More
miR-185-5p Regulates Inflammation and Phagocytosis through CDC42/JNK Pathway in Macrophages.
Authors:Ma X,Liu H,Zhu J,Zhang C,Peng Y,Mao Z,Jing Y,Chen F
Journal:Genes
PubMed ID:35328023
Macrophage activation is an essential component of systemic chronic inflammation and chronic inflammatory diseases. Emerging evidence implicates miR-185-5p in chronic inflammation diseases. However, the regulatory role of miR-185-5p in macrophage pro-inflammatory activation has not been studied previously. Here, we identified that miR-185-5p was one of the top genes and effectively ... More
Fetal Therapy Model of Myelomeningocele with Three-Dimensional Skin Using Amniotic Fluid Cell-Derived Induced Pluripotent Stem Cells.
Authors:Kajiwara K,Tanemoto T,Wada S,Karibe J,Ihara N,Ikemoto Y,Kawasaki T,Oishi Y,Samura O,Okamura K,Takada S,Akutsu H,Sago H,Okamoto A,Umezawa A
Journal:Stem cell reports
PubMed ID:28591652
Myelomeningocele (MMC) is a congenital disease without genetic abnormalities. Neurological symptoms are irreversibly impaired after birth, and no effective treatment has been reported to date. Only surgical repairs have been reported so far. In this study, we performed antenatal treatment of MMC with an artificial skin using induced pluripotent stem ... More
Turmeric Bioactive Compounds Alleviate Spinal Nerve Ligation-Induced Neuropathic Pain by Suppressing Glial Activation and Improving Mitochondrial Function in Spinal Cord and Amygdala.
Authors:Santos JM,Wang R,Bhakta V,Driver Z,Vadim Y,Kiritoshi T,Ji G,Neugebauer V,Shen CL
Journal:Nutrients
PubMed ID:37892476
This study examined the effects of turmeric bioactive compounds, curcumin C3 complex® (CUR) and bisdemethoxycurcumin (BDMC), on mechanical hypersensitivity and the gene expression of markers for glial activation, mitochondrial function, and oxidative stress in the spinal cord and amygdala of rats with neuropathic pain (NP). Twenty-four animals were randomly assigned ... More
Association between SNPs in microRNAs and microRNAs-Machinery Genes with Susceptibility of Leprosy in the Amazon Population.
Authors:da Silva MNS,da Veiga Borges Leal DF,Sena C,Pinto P,Gobbo AR,da Silva MB,Salgado CG,Dos Santos NPC,Dos Santos SEB
Journal:International journal of molecular sciences
PubMed ID:36142557
Leprosy is a chronic neurodermatological disease caused by the bacillus Mycobacterium leprae. Recent studies show that SNPs in genes related to miRNAs have been associated with several diseases in different populations. This study aimed to evaluate the association of twenty-five SNPs in genes encoding miRNAs related to biological processes and ... More