Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EIF2S1 Antibodies

          Invitrogen

          Phospho-EIF2S1 (Ser52) Polyclonal Antibody

          70 Published Figures
          128 References
          View all (56) EIF2S1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-EIF2S1 (Ser52) Polyclonal Antibody

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (70)
          Phospho-EIF2S1 (Ser52) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-EIF2S1 (Ser52) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 71

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-EIF2S1 (Ser52) Antibody (44-728G) in ICC/IF

          Rabbit anti eIF2alpha (pS52) phosphospecific polyclonal antibody. Left: Image shows immunocytochemical analysis of eIF2alpha (pS52) in rat brain (mediodorsal neocortex) sections following traumatic axonal injury (TAI) (24 h). Control (prior to injury induction) shows low basal levels of eIF2alpha (pS52) (see inset). Right: Image shows double labeling of APP (red) and eIF2alpha (pS52) (green) in neurons subj... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-EIF2S1 (Ser52) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)
          Phospho-EIF2S1 (Ser52) Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-EIF2S1 (Ser52) Polyclonal Antibody

          Product Details

          44-728G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 114 publications 114 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 2 publications 2 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          Inhibition Assays (Inhibition)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 6 publications 6 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Yeast

          Published species

          C. elegans, Cat, Chicken, Fruit fly, Human, Mouse, Non-human primate, Protozoa, Rat, Tag, Virus, Yeast

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human eIF2a that contains serine 52. This region is conserved among many species including rat, pig, cow, fruit fly and yeast.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533736

          Target Information

          EIF2a (Eukaryotic translation initiation factor 2A) is a heterotrimer composed of three subunits (alpha, beta, and gamma). This translation initiation factor drives binding of initiator methionyl-tRNA to the 40S ribosome in an AUG-dependent manner to form the 43S pre-initiation complex. The polypeptide can be phosphorylated by related protein kinases activated in response to stress. Phosphorylated EIF2A inhibits EIF2B activity and prevents guanine nucleotide exchange.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: eIF-2-alpha; eIF-2A; eIF-2alpha; eIF2 alpha; eIF2-alpha; eukaryotic translation initiation factor 2 subunit alpha; eukaryotic translation initiation factor 2, subunit 1 alpha, 35kDa; eukaryotic translation initiation factor 2A; translational initiation factor eIF-2, alpha subunit; unnamed protein product

          View more View less

          Gene Aliases: 0910001O23Rik; 2410026C18Rik; 35kDa; EIF-2; EIF-2A; EIF-2alpha; EIF2; EIF2A; eIF2alpha

          View more View less

          UniProt ID: (Human) P05198, (Mouse) Q6ZWX6

          View more View less

          Entrez Gene ID: (Human) 1965, (Mouse) 13665

          View more View less

          Function(s)
          nucleic acid binding RNA binding translation initiation factor activity protein binding ribosome binding cap-dependent translation initiation factor activity IRES-mediated translation initiation factor activity
          Process(es)
          mitophagy translation translational initiation regulation of translational initiation spermatogonial cell division negative regulation of translational initiation in response to stress stress granule assembly cellular response to amino acid starvation cellular response to oxidative stress cellular response to heat cellular response to UV response to endoplasmic reticulum stress regulation of translation in response to endoplasmic reticulum stress PERK-mediated unfolded protein response regulation of translational initiation in response to stress protein autophosphorylation cellular response to chemical stress mitochondrial respiratory chain supercomplex assembly HRI-mediated signaling response to kainic acid negative regulation of guanyl-nucleotide exchange factor activity response to manganese-induced endoplasmic reticulum stress positive regulation of type B pancreatic cell apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-nnlvt:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0