Customer Notice: Please be sure to follow the primer concentration guidelines as specified in the User Guide. For example, if you are using SYBR® Select with a StepOne™ or StepOnePlus™ Real-Time PCR Instrument under Standard Mode, please use a primer concentration less than 200 nM. Also, for these two instruments, please note that new StepOne™ Software v2.3 is also available for download.
SYBR™ Select Master Mix
Actual product may vary
Applied Biosystems™

SYBR™ Select Master Mix

SYBR™ Select Master Mix is a superior and cost-effective solution for your real-time PCR applications.Features of the SYBR™ Select MasterRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
447292010 x 5 mL
44729031 x 1 mL
44729081 x 5 mL
44729182 x 5 mL
44729195 x 5 mL
44729131 x 50 mL
Catalog number 4472920
Price (USD)
4,417.65
Online exclusive
4,855.00
Save 437.35
Each
Quantity:
10 x 5 mL
Price (USD)
4,417.65
Online exclusive
4,855.00
Save 437.35
Each
SYBR™ Select Master Mix is a superior and cost-effective solution for your real-time PCR applications.

Features of the SYBR™ Select Master Mix include:

• Specific—minimize primer-dimer and non-specific amplification
• Reproducible and sensitive—consistent amplification across a wide dynamic range
• Bright—contains SYBR™ GreenER™ dye for maximum brightness
• Carry-over contamination control—contains heat-labile UDG
• Can be used in either standard or fast cycling mode

Formulated for Maximum Specificity
SYBR™ Select Master Mix contains all the components needed for your real-time PCR reaction, except the template and primers, in a convenient 2X concentration premix. The master mix includes AmpliTaq™ DNA Polymerase, UP, a highly purified DNA polymerase with a proprietary hot start mechanism that provides exceptional specificity.

Obtain Reproducible Results Across a Wide Dynamic Range
SYBR Select Master Mix is specially formulated to provide robust results at 100 ng to 0.1 pg cDNA per reaction (Figure 1) and can reliably detect a single copy of genomic DNA (Figure 2).

Contains SYBR™ GreenER™ Dye and Heat-labile UDG
• SYBR™ GreenER™ dye is less inhibitory to PCR than SYBR™ Green I dye resulting in brighter signals
• Heat-labile uracil-DNA glycosylase (UDG) is included for worry-free carryover contamination control

Instrument Compatibility
SYBR™ Select Master Mix can be used in either standard or fast cycling mode and is compatible with all Applied Biosystems™ real-time PCR instruments except the 7900HT real-time PCR system. It is also compatible with the Bio-Rad IQ™5, Roche LightCycler™ LC480, and Stratagene™ MX3005P™ systems.

View performance data.

Alternative product
Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of SYBR Select Master Mix and added additional capabilities for your gene expression analysis.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, Fast Mode, StepOne™, Standard Mode, StepOnePlus™, Fast Mode, StepOnePlus™, Standard Mode, ViiA™ 7 System, AB StepOnePlus™, Fast Mode, AB StepOne™, Standard Mode, AB 7500, Fast Mode, AB 7500, Standard Mode, AB StepOne™, Fast Mode, AB StepOnePlus™, Standard Mode
FormatVial
Hot StartBuilt-In Hot Start
No. of Reactions5000 Reactions
Passive Reference DyeROX (Pre-mixed)
PolymeraseAmpliTaq DNA Polymerase
Product LineSYBR
Product TypeReal Time PCR SYBR Master Mix
Purity or Quality GradeUP (Ultra Pure)
Quantity10 x 5 mL
Sample TypeDNA (Genomic), cDNA
Sufficient For100 Reactions of 20 μL
Volume10 x 5 mL
Concentration2X
Detection MethodSYBR
For Use With (Application)Gene Expression
FormLiquid
GC-Rich PCR PerformanceHigh
Label or DyeSYBR GreenER
PCR MethodqPCR
Reaction SpeedFast or Standard
Unit SizeEach
Contents & Storage
The 2x mix contains SYBR™ GreenER™ dye, AmpliTaq™ DNA Polymerase UP, dNTPs with dUTP/dTTP blend, heat-labile UDG, ROX passive reference dye and optimized buffer components. Contains 1 X 1 mL tube, sufficient for 100, 20 μL reactions (10 μL Master Mix per reaction). Store at 2–8°C.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).

See user guide for primer concentration recommendations for the StepOne and StepOnePlus systems.

Frequently asked questions (FAQs)

What can I do to improve the sensitivity of my qPCR assay?

If you are targeting a low-abundance gene, you may have trouble getting Ct values in a good, reliable range (Ct > 32). To increase the sensitivity of the assay, you may want to consider the following:

- Increase the amount of RNA input into your reverse transcription reaction, if possible
- Increase the amount of cDNA in your qPCR reaction (20% by volume max)
- Try a different reverse transcription kit, such as our SuperScript VILO Master Mix, for the highest cDNA yield possible
- Consider trying a one-step or Cells-to-CT type workflow (depending on your sample type)

How do I set the baseline for my qPCR experiment?

Most times your instrument software can automatically set a proper baseline for your data. Check out our short video, Understanding Baselines, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=5BjFAJHW-bE).

How do I set the threshold for my qPCR experiment?

In most cases your instrument software can automatically set a proper threshold for your data. Check out our short video, Understanding Thresholds, for more information on how to set them (https://www.youtube.com/watch?feature=player_embedded&v=H_xsuRQIM9M).

I am not getting any amplification with my TaqMan Assay or SYBR Green primer set. What is causing this?

There could be several reasons for no amplification from an assay or primer set. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/no-amplification.html) for more details.

I am getting amplification in my no-template control (NTC) wells in my qPCR experiment. What is causing this?

There could be several reasons for amplification in a NTC well. Please see these examples and suggested solutions in our Real-Time Troubleshooting Tool (https://www.thermofisher.com/us/en/home/life-science/pcr/real-time-pcr/qpcr-education/real-time-pcr-troubleshooting-tool/gene-expression-quantitation-troubleshooting/amplification-no-template-control.html) for more details.