Ion Xpress™ Barcode Adapters 17-32 Kit - FAQs
Ion Xpress™ Barcode Adapters 17-32 Kit - FAQs
View additional product information for Ion Xpress™ Barcode Adapters 17-32 Kit - FAQs (4474009)
4 product FAQs found
Do libraries constructed with the Ion Xpress Barcode adapters require balancing to help ensure equal representation on the Ion PGM or Proton instrument sequencing run?
No, the Ion Xpress Barcode adapters are designed with a 3 bp adapter sequence (GAT) at the 3' end of each barcode to help minimize adapter ligation bias. When pooling barcoded libraries, accurate library quantification is extremely important to help ensure even barcode representation on the sequencing run.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many fragment or amplicon libraries can be made per barcode with the Ion Xpress Barcode Adapters kits?
The Ion Xpress Barcode Adapters kits are available as a set of 96 (Cat. No. 4474517) or in sets of 16 (Cat. Nos.4471250, 4474009, 4474518, 4474519, 4474520 or 4474521). Each barcode kit is sufficient for preparing ? 10 libraries per barcode (10 x 16 libraries or 10 x 96 libraries) for 100 ng input, or 2 libraries per barcode for 1 µg input.
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
How many samples can I multiplex on a single chip for sequencing on the Ion PGM or Proton System?
The number of samples that can be multiplexed in a single sequencing run depends on the capacity of the chip, the size of the library, and the required coverage. A table of approximate capacities based on the size of the library and chip can be found in the Ion Ampliseq Preparation User Guide (https://tools.thermofisher.com/content/sfs/manuals/MAN0006735_AmpliSeq_DNA_RNA_LibPrep_UG.pdf) within the section “Strategies for combining Ion Ampliseq libraries.”
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
I am planning to use the Oncomine BRCA Research Assay, Manual Library Preparation (Cat. No. A32840). Which other products would need I to order?
Here are the products you would need to order:
- Ion Xpress Barcode Adapters Kits
- Agencourt AMPure XP (Beckman Coulter Cat. No. A63880 or A63881)
- Ion Library Equalizer Kit (Cat. No. 4482298)