Product Image
Applied Biosystems™
Variant Reporter™ Software v2.0, Initial License
This is the Initial License for the Variant Reporter™ Software. This software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing, and resequencing with DNA sequencing files.
Have Questions?
Catalog NumberQuantity
44750061 License
Catalog number 4475006
Price (USD)
-
Quantity:
1 License
This is the Initial License for the Variant Reporter™ Software. This software performs comparative sequencing, also known as direct sequencing, medical sequencing, PCR sequencing, and resequencing with DNA sequencing files. The software is designed for reference-based and non-reference-based analysis such as mutation detection and analysis, SNP discovery and validation, and sequence confirmation. The robust algorithms will call SNPs, mutations, insertions, deletions, and heterozygous insertions/deletions for data generated using the Applied Biosystems™ genetic analyzers.
Related Links:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Data EntryCE Sequencing (.ab1 files)
For Use With (Application)Sequencing
LicenseInitial
Operating SystemWindows 7 Pro Service Pack 1
Product LineVariant Reporter
Product TypeSoftware V2.0, Initial License
Quantity1 License
SoftwareSequencing Data Analysis
Unit SizeEach
Contents & Storage
Software/CD
Frequently asked questions (FAQs)
We are upgrading to the Windows 10 operating system. What are the compatible capillary electrophoresis secondary analysis software with Windows 10?
The following capillary electrophoresis secondary analysis software are compatible with Windows 10 (and also Windows 7):
-Sequencing Analysis Software v7
-SeqScape Software v4
-Variant Reporter Software v3
-GeneMapper Software v6
-Minor Variant Finder v1.2
-MicrobeBridge v1.1
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where do I get the latest Applied Biosystems Capillary Electrophoresis Software patches?
Please visit our Applied Biosystems Genetic Analyzer Freeware and Patches website: https://www.thermofisher.com/sangerpatches
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I import my data from Variant Reporter Software?
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I archive a selected project in the Variant Reporter Software?
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up the Data Store in the Variant Reporter Software?
To back up the Data Store:
1. Open the Dashboard View.
2. Click .
3. In the Browse for Back-up Folder Destination dialog box, select the folder that you want to back up the Data Store to, then click OK. Your project will start to get backed up.
Note: All associated Projects, Analysis Parameters, and References contained in the current Data Store will be copied to the folder you selected when you back up the Variant Reporter Data Store.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.