Variant Reporter™ Software v2.0, Initial License - FAQs
Variant Reporter™ Software v2.0, Initial License - FAQs
View additional product information for Variant Reporter™ Software v2.0, Initial License - FAQs (4475006)
10 product FAQs found
We are upgrading to the Windows 10 operating system. What are the compatible capillary electrophoresis secondary analysis software with Windows 10?
The following capillary electrophoresis secondary analysis software are compatible with Windows 10 (and also Windows 7):
-Sequencing Analysis Software v7
-SeqScape Software v4
-Variant Reporter Software v3
-GeneMapper Software v6
-Minor Variant Finder v1.2
-MicrobeBridge v1.1
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Where do I get the latest Applied Biosystems Capillary Electrophoresis Software patches?
Please visit our Applied Biosystems Genetic Analyzer Freeware and Patches website: https://www.thermofisher.com/sangerpatches
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I import my data from Variant Reporter Software?
For importing data, there are many ways to do so:
More information on backing up the Data Store can be found in the Variant Reporter Help Contents under the keyword “Back-up”.
1. Select the Data Store where you would like the data imported to. To choose a Data Store:
a. Click the Manage button in the Data Store section of the Dashboard View.
b. Highlight the Data Store you wish to use and click on Change Location to select it. Click Close to close the window.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I archive a selected project in the Variant Reporter Software?
To archive a Variant Reporter software project from the Dashboard:
1. Open the Dashboard View.
2. In the Project tab select the project you want to archive, then click .
3. In the Archive Destination dialog box, browse to the folder where you want the project archived.
4. Click OK. The project file you archived will no longer appear in the Variant Reporter Dashboard.
More information on archiving Project data can be found in the Variant Reporter Help Contents under the keyword “Archive”.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up the Data Store in the Variant Reporter Software?
To back up the Data Store:
1. Open the Dashboard View.
2. Click .
3. In the Browse for Back-up Folder Destination dialog box, select the folder that you want to back up the Data Store to, then click OK. Your project will start to get backed up.
Note: All associated Projects, Analysis Parameters, and References contained in the current Data Store will be copied to the folder you selected when you back up the Variant Reporter Data Store.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I export a Project, Analysis Parameter, or Reference from the Dashboard?
More information on exporting data can be found in the Variant Reporter Help Contents under the keyword “Export”.
To export a Project, Analysis Parameter, or Reference from the Dashboard:
Open the Dashboard View (see below), then click on the Tab (Projects, Analysis Parameters, or References) where the file you want to export is located and left-click on the file name to select it.
Click Export to open the Export File dialog box.
Select the location you want to save the file in, then click Save.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
How can I back up my Variant Reporter Software data?
For exporting and backing up the data, there are many ways to do so:
Export: takes the Project, Analysis Parameters, and Reference Data of your choice and saves them to a location of your choice. You can use this option for backing up data or sending data to support personnel. Exported files will not be removed from the Dashboard unless you delete them.
Archive: for Projects only: will take a selected Project and export it to a location of your choice. The Project is then removed from the Project Dashboard. You cannot archive Analysis Parameters or Reference Data.
Back-up: will duplicate all of the information in the Data Store (Projects, Analysis Parameters, and Reference Data) to a folder of your choice. This is an all-or-nothing option - you cannot select specific files for back-up. If you wish to have specific files backed up, please use the Export option.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We just upgraded to Variant Reporter Software v.2 and I'm wondering if my old data files will work on it?
The Variant Reporter Software v.2 is compatible with the following data files:
-3130 series Data Collection Software 4
-3730 series Data Collection Software 4
-3500 series Data Collection Software 20
For more information, please click here for the Variant Reporter Software v.2 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Variant-Reporter-Software-v-2-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
I have a copy of Variant Reporter Software v.2 and want to know what the computer requirements are.
The computer requirements are as follows:
-Minimum: 2.0 GHz or faster Intel processor
-2 GB RAM
-1 GB hard disk space for application
Recommended CPU: Core 2 Duo E8400/3.0 GHz
-4 GB RAM
-1 GB hard disk space
For the operating system, Variant Reporter Software v.2 requires Windows 7 Professional, 32-bit edition, service pack 1. Please click here for the Variant Reporter Software v.2 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Variant-Reporter-Software-v-2-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
We are upgrading to the Windows 7 operating system. Can I install Variant Reporter Software v.2 on that?
This software has been extensively tested on Windows 7 SP1 (32-bit version). While there are some indications that the software will run normally on the 64-bit version of Windows 7 SP1 operating system, the 64-bit configuration is not officially supported. It is strongly recommended that this software is run on Windows 7 SP1 (32-bit).
If installing on the instrument computer, the update will require a new computer and upgrading the Data Collection Software as well. Please click here for the Variant Reporter Software v.2 datasheet (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Variant-Reporter-Software-v-2-datasheet.pdf).
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.