Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
TaqMan®  OpenArray® Human  Kinome Panel
Actual product may vary
Applied Biosystems™
TaqMan® OpenArray® Human Kinome Panel
Have Questions?
Catalog number 4475368
Price (USD)
1,058.00
1 each
Price (USD)
1,058.00
1 each
The TaqMan® OpenArray® Human Kinome Panel is an efficient, easy-to-use OpenArray® plate for the characterization of genes encoding human kinases. This panel targets 772 well-defined genes, plus 56 endogenous control genes. The format of the OpenArray® plate allows for 1–3 replicates to be run in parallel per plate.
The TaqMan® OpenArray® Human Kinome Panel is intended to capture the full set of protein kinases in the human genome. Genes were chosen based on the primary literature and public databases. In addition, there are 56 endogenous control genes against which the assays can be normalized. All assays in this panel have been bench-validated in our laboratories, so you can be sure that each assay is fully optimized.
The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.
This panel is intended for use with the TaqMan® OpenArray® Real-time PCR System, which can cycle and image 3 plates simultaneously, providing high-throughput results with ease.
Note: equivalent panels for use with the QuantStudio™ 12K Flex instrument are also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 each
Contents & Storage
Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C