Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
TaqMan®  OpenArray® Human Stem Cell Panel
Actual product may vary
Applied Biosystems™
TaqMan® OpenArray® Human Stem Cell Panel
Have Questions?
Catalog number 4475370
Price (USD)
1,058.00
1 each
Price (USD)
1,058.00
1 each
The TaqMan® OpenArray® Human Stem Cell Panel is an efficient, easy-to-use OpenArray® plate for the characterization of undifferentiated stem cells or their differentiated derivatives. This gene signature panel targets 609 well-defined genes validated as markers for pluripotency, plus 22 endogenous control genes. The format of the OpenArray® plate allows for 1–4 replicates to be run in parallel per plate.
The TaqMan® OpenArray® Human Stem Cell Panel contains a well-defined set of validated gene expression markers to characterize human embryonic stem (ES) cell identity and assess phenotypic variations between embryonic stem cell isolates. The gene content of this OpenArray® plate resulted directly from the work of the I.S.C.I consortium and characterization of human embryonic stem cell lines by the International Stem Cell Initiative.
The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.
The Right Targets
This OpenArray® plate uses the proprietary Applied Biosystems® OpenArray® technology to simultaneously analyze gene expression levels by quantitative PCR for 609 test genes and 22 endogenous controls.
Genes were included based on the following criteria:
• Expression in human embryonal carcinoma cells (hEC), primordial germ cells, or human ES (hES) cells
• Known role in maintenance of pluripotency in hES cells
• Identification in microplate-based experiments as being correlated with the stem cell state
• Expression known to be characteristic of specific differentiation lineages in the post implantation embryo and mature tissues
This panel is intended for use with the TaqMan® OpenArray® Real-time PCR System, which can cycle and image 3 plates simultaneously, providing high-throughput results with ease.
Note: equivalent panels for use with the QuantStudio™ 12K Flex instrument are also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 each
Contents & Storage
Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C