Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
TaqMan®  OpenArray® Human Cancer Panel
Actual product may vary
Applied Biosystems™
TaqMan® OpenArray® Human Cancer Panel
Have Questions?
Catalog number 4475371
Price (USD)
1,058.00
1 each
Price (USD)
1,058.00
1 each
The TaqMan® OpenArray® Human Cancer Panel is an efficient, easy-to-use OpenArray® plate for the characterization of undifferentiated cancers or their differentiated derivatives. This gene signature panel targets 624 well-defined genes validated as markers for pluripotency, plus 24 endogenous controls. The format of the OpenArray® plate allows for 1–4 replicates to be run in parallel per plate.
The TaqMan® OpenArray® Human Cancer Panel targets genes related to DNA repair, angiogenesis, cell adhesion, and ECM, as well as genes involved in the cell cycle and apoptosis, and many of the genes encoding kinases and transcription factors that have been found to be differentially expressed in early cancer and metastatic disease. In addition, 24 endogenous control genes were included against which the assays can be normalized. All assays in this panel were developed using a high level of bioinformatics and wet-lab stringency.
The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.
This panel is intended for use with the TaqMan® OpenArray® Real-time PCR System, which can cycle and image 3 plates simultaneously, providing high-throughput results with ease.
Note: equivalent panels for use with the QuantStudio™ 12K Flex instrument are also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 each
Contents & Storage
Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C