Not what you're looking for? Search all of our TaqMan® arrays for your gene or pathway of interest.
TaqMan®  OpenArray® Mouse Inflammation Panel
Actual product may vary
Applied Biosystems™
TaqMan® OpenArray® Mouse Inflammation Panel
Have Questions?
Catalog number 4475373
Price (USD)
1,058.00
1 each
Price (USD)
1,058.00
1 each
The TaqMan® OpenArray® Mouse Inflammation Panel is an efficient, easy-to-use OpenArray® plate for quantitative gene expression analysis of inflammation genes important in drug discovery.
The panel covers 632 genes that have been studied as targets for a range of inflammatory diseases, plus 16 endogenous control genes. The plate is pre-formatted and inventoried, cost-effective, convenient, and easy-to-use, without the need for expensive robotics. Results are reproducible and consistent across samples, studies, and labs—for the same data quality from plate-to-plate and lot-to-lot, even with different operators.
This panel is intended for use with the TaqMan® OpenArray® Real-time PCR System, which can cycle and image 3 plates simultaneously, providing high-throughput results with ease.
Note: equivalent panels for use with the QuantStudio™ 12K Flex instrument are also available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Unit Size1 each
Contents & Storage
Includes one OpenArray® Plate containing a defined panel of pre-loaded, bench-validated TaqMan® assays.
Store at -20 °C