Are there Ion Torrent fragment library kits available for use with my AB Library Builder System?
The currently available kits for the AB Library Builder System instrument includes:
- Ion Plus Library Kit for AB Library Builder System (Cat. No. 4477597)
- Ion Xpress Plus Library Kit for AB Library Builder System (Cat. No. 4477598).
Both kits require the use of the separately available Ion Plus Fragment Library Adapters Kit (Cat. No. 4476340) or the Ion Xpress Barcode Adapters kits and the Ion Plus and Ion Xpress Plus Protocol Card for AB Library Builder System (Cat. No. 4477687).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.
- Ion Plus Library Kit for AB Library Builder System (Cat. No. 4477597)
- Ion Xpress Plus Library Kit for AB Library Builder System (Cat. No. 4477598).
Both kits require the use of the separately available Ion Plus Fragment Library Adapters Kit (Cat. No. 4476340) or the Ion Xpress Barcode Adapters kits and the Ion Plus and Ion Xpress Plus Protocol Card for AB Library Builder System (Cat. No. 4477687).
Find additional tips, troubleshooting help, and resources within our Next-Generation Sequencing Support Center.