Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EGFR Antibodies

          Invitrogen

          Phospho-EGFR (Tyr1173) Polyclonal Antibody

          Advanced Verification
          13 Published Figures
          30 References
          View all (354) EGFR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-EGFR (Tyr1173) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (4)
          • Published Figures (13)
          • Advanced Verification (3)
          Phospho-EGFR (Tyr1173) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-EGFR (Tyr1173) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 20

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-EGFR (Tyr1173) Antibody (44-794G) in ICC/IF

          Immunofluorescence analysis of EGFR (pY1173) was done on 70% confluent log phase HeLa cells treated with EGF (200 ng/mL for 10 min). The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with EGFR (pY1173) Rabbit polyclonal Antibody (Product # 44-794G) at 2 µg/mL in 1... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-EGFR (Tyr1173) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-EGFR (Tyr1173) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-EGFR (Tyr1173) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-EGFR (Tyr1173) Antibody
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody in Western Blot (WB)
          Phospho-EGFR (Tyr1173) Antibody
          Phospho-EGFR (Tyr1173) Antibody
          Phospho-EGFR (Tyr1173) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-EGFR (Tyr1173) Polyclonal Antibody

          Product Details

          44-794G

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 22 publications 22 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          View 2 publications 2 publications

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Bacteria, Human, Mouse, Sheep

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human EGFR that contains tyrosine 1173. The sequence is conserved in mouse and rat.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Antigen affinity chromatography

          Storage buffer

          Dulbecco's PBS, pH 7.3, with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533757

          Product Specific Information

          44-794G has been used successfully in peptide-ELISA. Dilution is to be determined by the consumer.

          Target Information

          EGFR, epidermal growth factor receptor, is a receptor tyrosine kinases that signals in response to various growth factors. Overexpression has been linked to numerous types of cancer and EGFR is the target of both biological and small molecular therapeutics. EGFR is encoded by the EGFR gene located on chromosome 7 in humans. EGFR belongs to the HER/ERbB family of proteins that includes three other receptor tyrosine kinases, ERbB2, ERbB3, ERbB4. EGFR is a transmembrane receptor and binding of its cognate ligands such as EGF (Epidermal Growth Factor) and TGF alpha (Transforming Growth Factor alpha) to the extracellular domain leads to EGFR dimerization followed by autophosphorylation of the tyrosine residues in the cytoplasmic domain. Overexpression is observed in tumors of the head and neck, brain, bladder, stomach, breast, lung, endometrium, cervix, vulva, ovary, esophagus, stomach and in squamous cell carcinoma.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 2.7.10.1; avian erythroblastic leukemia viral (v-erb-b) oncogene homolog; avian erythroblastic leukemia viral (v-erbB) oncogene homolog; cell growth inhibiting protein 40; cell proliferation-inducing protein 61; contains extracellular domain; EC 2.7.10.1; EGFR-related peptide; Epidermal growth factor receptor formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); epidermal growth factor receptor tyrosine kinase domain; epidermal growth factor receptor, formerly avian erythroblastic leukemia viral (v-erbB) oncogene homolog (Erbb1); erb-b2 receptor tyrosine kinase 1; Erbb1; GIG40; kinase EGFR; membrane protein; Oncogene ERBB; PIG61; proto-oncogene c-ErbB-1; receptor tyrosine-protein kinase erbB-1; tyrosine protein kinase; unnamed protein product; Urogastrone; waved 2

          View more View less

          Gene Aliases: 9030024J15Rik; ERBB; ErbB-1; ERBB1; Errb1; ERRP; HER1; mENA; NISBD2; NNCIS; PIG61; wa-2; wa2; Wa5

          View more View less

          UniProt ID: (Human) P00533, (Mouse) Q01279

          View more View less

          Entrez Gene ID: (Human) 1956, (Rat) 24329, (Mouse) 13649

          View more View less

          Function(s)
          chromatin binding double-stranded DNA binding protein kinase activity MAP kinase kinase kinase activity protein tyrosine kinase activity transmembrane receptor protein tyrosine kinase activity transmembrane signaling receptor activity epidermal growth factor-activated receptor activity protein binding ATP binding kinase activity enzyme binding kinase binding protein phosphatase binding protein tyrosine kinase activator activity ubiquitin protein ligase binding identical protein binding cadherin binding epidermal growth factor binding actin filament binding ATPase binding nucleotide binding receptor binding integrin binding calmodulin binding protein kinase binding macromolecular complex binding
          Process(es)
          cell morphogenesis liver development embryonic placenta development positive regulation of protein phosphorylation hair follicle development translation receptor-mediated endocytosis response to oxidative stress signal transduction cell surface receptor signaling pathway transmembrane receptor protein tyrosine kinase signaling pathway epidermal growth factor receptor signaling pathway salivary gland morphogenesis midgut development circadian rhythm positive regulation of cell proliferation epidermis development gene expression diterpenoid metabolic process peptidyl-tyrosine phosphorylation telencephalon development cerebral cortex cell migration neuron differentiation positive regulation of cell growth lung development positive regulation of cell migration response to estradiol positive regulation of superoxide anion generation positive regulation of peptidyl-serine phosphorylation response to cobalamin response to hydroxyisoflavone response to lipid positive regulation of toll-like receptor 9 signaling pathway intracellular signal transduction ERBB2-EGFR signaling pathway negative regulation of epidermal growth factor receptor signaling pathway regulation of cell proliferation negative regulation of protein catabolic process vasodilation positive regulation of phosphorylation ovulation cycle hydrogen peroxide metabolic process xenobiotic transport negative regulation of apoptotic process positive regulation of MAPK cascade protein kinase B signaling tongue development positive regulation of DNA repair positive regulation of DNA replication positive regulation of bone resorption positive regulation of vasoconstriction negative regulation of mitotic cell cycle positive regulation of transcription from RNA polymerase II promoter protein autophosphorylation astrocyte activation positive regulation of fibroblast proliferation digestive tract morphogenesis positive regulation of smooth muscle cell proliferation neuron projection morphogenesis positive regulation of epithelial cell proliferation regulation of peptidyl-tyrosine phosphorylation response to calcium ion positive regulation of protein kinase B signaling positive regulation of synaptic transmission, glutamatergic positive regulation of glial cell proliferation morphogenesis of an epithelial fold eyelid development in camera-type eye response to UV-A positive regulation of mucus secretion positive regulation of ERK1 and ERK2 cascade cellular response to amino acid stimulus cellular response to mechanical stimulus cellular response to growth factor stimulus cellular response to estradiol stimulus cellular response to xenobiotic stimulus cellular response to dexamethasone stimulus positive regulation of protein kinase C signaling positive regulation of canonical Wnt signaling pathway liver regeneration cell-cell adhesion positive regulation of G1/S transition of mitotic cell cycle positive regulation of prolactin secretion positive regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of protein localization to plasma membrane negative regulation of cardiocyte differentiation learning or memory negative regulation of macroautophagy cellular response to epidermal growth factor stimulus positive regulation of cGAS/STING signaling pathway positive regulation of protein localization to early endosome MAPK cascade ossification protein insertion into membrane
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch