MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates with Barcode, 20 Plates - FAQs
MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates with Barcode, 20 Plates - FAQs
View additional product information for MicroAmp™ EnduraPlate™ Optical 96-Well Clear Reaction Plates with Barcode - FAQs (4483352, 4483354)
1 product FAQs found
What is included in Oncomine Precision Assay GX (Cat. No. A46291)?
Oncomine Precision Assay GX (Cat. No. A46291) includes the following:
- OPA Pool 1 panel (Part No. A44350)
- Genexus Strip 1, quantity of 8 strips (Part No. A46812)
- Genexus Strip 2-HD, quantity of 8 strips (Part No. A46814)
- The OPA Pool 1 is provided in 8 pairs of tubes, where each tube pair contains one tube with forward (FWD) primers and one tube with reverse (REV) primers.