The TaqMan™ assay provides a simple, reliable, and rapid method for detecting the presence of a specific bacterial or viralRead more
Have Questions?
Catalog Number
Quantity
4485059
96 reactions
Catalog number 4485059
Price (USD)
1,616.00
96 reactions
Quantity:
96 reactions
Price (USD)
1,616.00
96 reactions
The TaqMan™ assay provides a simple, reliable, and rapid method for detecting the presence of a specific bacterial or viral target. The assay uses PCR to amplify a DNA target unique to the microorganism and TaqMan™ probe to detect and quantify the target, all in an an easy-to-use, lyophilized format. TaqMan™ assays deliver the fastest RT-PCR run times on the market (<40 minutes in Fast mode). Simply add 30 μL of purified DNA to the assay tube and run on Fast or standard instrumentation. The TaqMan™ assays provide microbial detection capabilities and high-confidence results to help accelerate your food and environmental testing needs.
Features of the TaqMan™ assays:
• Convenience—simply add your nucleic acid sample to the assay tube and run; no pre-mixing or master mix preparation required • Consistency—fewer operator steps means reduced error and increased reproducibility across time, users, and laboratories • Improved data accuracy—an internal positive control is included to monitor your PCR • Fast chemistry—40-min run time with Fast cycling, compatible with standard or Fast modes
TaqMan™ assays are suitable for single or multiplex analysis. Because they are run using universal conditions, multiple assays can be analyzed in the same run. The robust assay design provides highly accurate, reproducible, and reliable results, with the enhanced stability of a lyophilized format. A passive internal reference based on proprietary ROX™ dye is included for precise data analysis.
This product is for environmental testing, quality control/quality assurance testing, food and agricultural testing, and research purposes only.
Custom assays have been bioinformatically developed and tested for performance. Validation services for specific samples types are provided by our Custom Solutions team. Please contact your local Thermo Fisher Scientific representative for more information.
Specifications
DescriptionIncludes Master Mix
No. of Reactions96 reactions
Quantity96 reactions
SpeciesAll Species
Sufficient For96 Reactions
Target Organism ClassE. coli
Detection MethodPrimer-probe
For Use With (Application)Food and Beverage
FormLyophilized
Reaction SpeedFast or Standard
Unit Size96 reactions
Contents & Storage
• 12 8-tube strips (store at 2–8°C in the dark) • Optical 8-tube strips and 8-cap strips (store at room temperature)