Beginning August 15, 2014, this product will ship under ambient conditions. Full details can be found here.
TaqMan™ GAPDH Assay, JUN™ dye/QSY™ probe, primer-limited
Applied Biosystems™

TaqMan™ GAPDH Assay, JUN™ dye/QSY™ probe, primer-limited

The Applied Biosystems™ TaqMan™ GAPDH Assay with JUN™ dye-labeled QSY™ probe (primer limited) provides a pre-formulated assay for quantitating humanRead more
Have Questions?
Catalog NumberQuantity
4485713250 reactions
Catalog number 4485713
Price (USD)
415.65
Online exclusive
442.00
Save 26.35
Each
Quantity:
250 reactions
Price (USD)
415.65
Online exclusive
442.00
Save 26.35
Each
The Applied Biosystems™ TaqMan™ GAPDH Assay with JUN™ dye-labeled QSY™ probe (primer limited) provides a pre-formulated assay for quantitating human GAPDH. This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. It consists of a JUN™ dye-labeled QSY™ probe plus sequence-specific forward and reverse primers. The assay can be used for multiplex or singleplex PCR reactions, and it is primer limited, so it can be used with samples that overexpress GAPDH.

Benefits of this assay include:

• Eliminate assay design time by using pre-designed primers and pre-designed JUN™ dye-labeled TaqMan™ probe
• Minimize experimental optimization time
• Use in conjunction with FAM™ or VIC™ dye-labeled predesigned gene expression assays
• Formulated at a primer-limited concentration for use in multiplex reactions

Save time in assay design, formulation, and testing by using the TaqMan™ GAPDH Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.

In order to facilitate multiplex experiments, the assay formulation is primer limited, allowing the assay to be used with high GAPDH expressors.

The JUN™ dye is optimized for use with the 4th filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments. Please note that master mix containing ROX™ dye cannot be used with this assay (with JUN™ dye) due to an overlap of spectra. TaqMan™ Multiplex Master Mix with MUSTANG PURPLE™ dye is recommended.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNM_002046.3
Amplicon Size118
FormatTube
Gene SymbolGAPDH
Internal Probe ModificationQSY™ Non-Fluorescent Quencher (3')
No. of Reactions250 Reactions
Product LineTaqMan
Product TypeGapdh Assay
Quantity250 reactions
SpeciesHuman
Sufficient For250 Reactions
TargetGlyceraldehyde-3-phosphate dehydrogenase
Detection MethodPrimer-probe
For Use With (Application)Gene Expression
GC-Rich PCR PerformanceHigh
Label or DyeJUN
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Primer-Probe Set(s)

Frequently asked questions (FAQs)

What are the excitation and emission profiles for ABY, JUN, Cyanine 5, and Cyanine 5.5 dyes?

ABY dye has an excitation wavelength of 568 nm and emission wavelength of 583 nm.
JUN dye has an excitation wavelength of 606 nm and emission wavelength of 618 nm.
Cynanine 5 dye has an excitation wavelength of 649 nm and emission wavelength of 667 nm.
Cyanine 5.5 dye has an excitation wavelength of 678 nm and emission wavelength of 695 nm.

Please refer to your instrument user guide for additional information on compatibility with different dye channels.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.