Beginning August 15, 2014, this product will ship under ambient conditions. Full details can be found here.
TaqMan™ RNase P Assay, ABY™ dye/QSY™ probe, primer-limited
Applied Biosystems™

TaqMan™ RNase P Assay, ABY™ dye/QSY™ probe, primer-limited

The Applied Biosystems™ TaqMan™ RNase P Assay with ABY™ dye-labeled QSY™ probe (primer limited) provides a pre-formulated assay for quantitatingRead more
Have Questions?
Catalog NumberQuantity
4485715250 reactions
Catalog number 4485715
Price (USD)
407.65
Online exclusive
434.00
Save 26.35
Each
Quantity:
250 reactions
Price (USD)
407.65
Online exclusive
434.00
Save 26.35
Each
The Applied Biosystems™ TaqMan™ RNase P Assay with ABY™ dye-labeled QSY™ probe (primer limited) provides a pre-formulated assay for quantitating human RNase P. This assay enables relative gene expression quantification in cDNA samples when used with other gene expression assays. It consists of an ABY™ dye-labeled QSY™ probe plus sequence-specific forward and reverse primers. The assay can be used for multiplex or singleplex PCR reactions, and it is primer limited, so it can be used with samples that overexpress RNase P.

Benefits of this assay include:

• Eliminate assay design time using pre-designed primers and pre-designed ABY™ dye-labeled TaqMan™ probe
• Minimize experimental optimization time
• Use in conjunction with FAM™ or VIC™ dye-labeled predesigned gene expression assays
• Formulated at a primer-limited concentration for use in multiplex reactions

Save time in assay design, formulation, and testing by using the TaqMan™ RNase P Assay as your control when quantifying gene expression. This pre-designed probe and primer set enables you to normalize the amount of sample RNA or DNA in a reaction when used with a control sample. Or use it for relative gene expression quantification in cDNA samples when used with other gene expression assays.

In order to facilitate multiplex experiments, the assay formulation is primer limited, allowing the assay to be used with high RNase P expressors.

The ABY™ dye is optimized for use with the 3rd filter of our QuantStudio™, ViiA™ 7, and 7500 series real-time PCR instruments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Accession NumberNR_002312.1
Amplicon Size87
FormatTube
Gene SymbolRNase P (RPPH1)
Internal Probe ModificationQSY™ Non-Fluorescent Quencher (3')
No. of Reactions250 reactions
Product LineTaqMan
Product TypeReal Time PCR Reagents
Quantity250 reactions
SpeciesHuman
TargetRibonuclease P RNA component H1
Detection MethodPrimer-probe
For Use With (Application)Real Time PCR (qPCR)
GC-Rich PCR PerformanceHigh
Label or DyeABY
PCR MethodqPCR
Unit SizeEach
Contents & Storage
Primer-Probe Set(s)

Frequently asked questions (FAQs)

What are the excitation and emission profiles for ABY, JUN, Cyanine 5, and Cyanine 5.5 dyes?

ABY dye has an excitation wavelength of 568 nm and emission wavelength of 583 nm.
JUN dye has an excitation wavelength of 606 nm and emission wavelength of 618 nm.
Cynanine 5 dye has an excitation wavelength of 649 nm and emission wavelength of 667 nm.
Cyanine 5.5 dye has an excitation wavelength of 678 nm and emission wavelength of 695 nm.

Please refer to your instrument user guide for additional information on compatibility with different dye channels.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.