VetMAX™ Mycoplasma Gallisepticum/Synoviae Reagent
Applied Biosystems™

VetMAX™ Mycoplasma Gallisepticum/Synoviae Reagent

VetMAX Mycoplasma gallisepticum/synoviae Reagent contains primers/probes targeting Mycoplasma gallisepticum/synoviae, an agent of upper respiratory infection in poultry. Infections of multiple-age layer flocks are common and may contribute to decreased egg production. The infection can be transmitted vertically and horizontally.
Have Questions?
Catalog NumberQuantity
4486959100 reactions
Catalog number 4486959
Price (USD)
900.00
Each
Quantity:
100 reactions
Price (USD)
900.00
Each

VetMAX Mycoplasma gallisepticum/synoviae Reagent contains primers/probes targeting Mycoplasma gallisepticum/synoviae, an agent of upper respiratory infection in poultry. Infections of multiple-age layer flocks are common and may contribute to decreased egg production. The infection can be transmitted vertically and horizontally. Once infected, individuals and flocks become chronic carriers. Real-time PCR is the most commonly used identification method for these mircroorganisms, enabling precise identification of pathogen nucleic acid and allowing for reliable and rapid testing of animal-derived samples.

VetMAX Mycoplasma gallisepticum/synoviae Reagent is approved for use as part of the NPIP (National Poultry Improvement Plan)-certified diagnostic test for Mycoplasma gallisepticum/synoviae as of 2016 under the United States voluntary control program.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
For Use With (Equipment)7500Fast, QuantStudio 5
No. of Reactions100 Reactions
Product LineVetMAX
Product TypePCR reagent for Animal Health
Quantity100 reactions
Sample TypePoultry samples
TargetM. synoviae (MS), M. gallisepticum (MG)
Target Organism ClassM. synoviae (MS), M. gallisepticum (MG)
Detection MethodPrimer-probe
Unit SizeEach
Contents & Storage
Store at -25°C to -15 °C.