TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR)
TaqMan™ PreAmp Master Mix pre-amplifies small amounts of cDNA without introducing amplification bias to the sample. Preamplification enables you toRead more
Have Questions?
Change view
Catalog Number
No. of Reactions
4488593
200 Reactions
4391128
40 Reactions
2 Options
Catalog number 4488593
Price (USD)
5,482.65
Online exclusive
6,160.00
Save 677.35
Each
No. of Reactions:
200 Reactions
Price (USD)
5,482.65
Online exclusive
6,160.00
Save 677.35
Each
TaqMan™ PreAmp Master Mix pre-amplifies small amounts of cDNA without introducing amplification bias to the sample. Preamplification enables you to stretch your precious sample into many more real-time PCR reactions.
TaqMan™ PreAmp Master Mix lets you:
• Analyze mRNA from precious samples such as laser capture microdissections, needle biopsies, and formalin-fixed paraffin-embedded (FFPE) tissues • Stretch as little as 1 ng of cDNA into 200 real-time PCR reactions for gene expression analysis using TaqMan™ Gene Expression assays • Analyze up to 100 gene expression targets with minimal hands-on time
Preserves Equilibrium of Targets Without Preamplification Bias In the past, preamplification kits and techniques often resulted in uneven amplification of targets and incorrect or biased data (as shown by low correlation coefficients between preamplified and unamplified samples). TaqMan™ PreAmp Master Mix amplifies with no bias and provides extremely high correlation between amplified and unamplified cDNA.
Easy Workflow With TaqMan™ PreAmp Master Mix, preamplification setup takes only a few minutes. After a one-time pooling of the TaqMan™ assays , preamplification setup (mixing the pooled assays with cDNA sample and TaqMan™ PreAmp Master Mix) takes only five additional minutes. Cycling takes 1.5 hours using a GeneAmp™ 9700 thermal cycler.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 Fast System, 7500 System, 7900HT System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7, StepOne™, StepOnePlus™, ViiA™ 7 System
FormatBottle
No. of Reactions200 Reactions
Product LineTaqMan
Product TypePreAmp Master Mix
Quantity200 reactions
Sample TypecDNA
Sufficient For200 Reactions
Volume5 mL
Detection MethodPrimer-probe
For Use With (Application)Pre-amplification
GC-Rich PCR PerformanceHigh
PCR MethodqPCR
Reaction SpeedStandard
Unit SizeEach
Contents & Storage
Contains one 5-mL bottle of 2X master mix.
Store at 2–8°C.
Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Frequently asked questions (FAQs)
Can I use preamplification with the TaqMan SNP Genotyping Assays?
There is a preamplification protocol included with our TaqMan Sample-to-SNP Kit. This is designed to work with a lysate sample.
How does the SuperScript IV Single Cell/Low Input cDNA PreAmp Kit differ from TaqMan PreAmp Master Mix?
TaqMan PreAmp Master Mix is designed for targeted preamplification of up to 100 targets from small amounts of cDNA, while SuperScript IV Single Cell/Low Input cDNA PreAmp Kit allows global preamplification of full-length cDNA from single cells or low input RNA. It provides reagents for cell lysis, reverse transcription and preamplification steps.
How do I reorder the TaqMan PreAmp Master Mix, Cat No. 4384266? It does not match the Cat. No. when I search online.
This is an internal part number for the TaqMan PreAmp Master Mix. To order, please use Cat No. 4391128 for the 40 reaction size or Cat. No. 4488593 for the 200 reaction size. This is the same product.