TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR), 200 reactions - FAQs
TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR), 200 reactions - FAQs
View additional product information for TaqMan™ PreAmp Master Mix (cDNA pre-amplification for qPCR) - FAQs (4488593, 4391128)
3 product FAQs found
Can I use preamplification with the TaqMan SNP Genotyping Assays?
There is a preamplification protocol included with our TaqMan Sample-to-SNP Kit. This is designed to work with a lysate sample.
How does the SuperScript IV Single Cell/Low Input cDNA PreAmp Kit differ from TaqMan PreAmp Master Mix?
TaqMan PreAmp Master Mix is designed for targeted preamplification of up to 100 targets from small amounts of cDNA, while SuperScript IV Single Cell/Low Input cDNA PreAmp Kit allows global preamplification of full-length cDNA from single cells or low input RNA. It provides reagents for cell lysis, reverse transcription and preamplification steps.
How do I reorder the TaqMan PreAmp Master Mix, Cat No. 4384266? It does not match the Cat. No. when I search online.
This is an internal part number for the TaqMan PreAmp Master Mix. To order, please use Cat No. 4391128 for the 40 reaction size or Cat. No. 4488593 for the 200 reaction size. This is the same product.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.